Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00474326_s1
          See other PCDHB9 GE Assays ›
          Gene Symbol:
          Pcdhb9
          Gene Name:
          protocadherin beta 9
          Gene Aliases:
          Pcdhb4C, PcdhbI
          Chromosome Location:
          Chr.18: 37400855 - 37403909 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00474326_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pcdhb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053134.3 NP_444364.3 1 1161 97
          GenBank mRNA AK032626.1 - 1 1162 97
          AY013791.1 - 1 956 97
          BC055319.1 - 1 1136 5703531 97
          BC062640.1 - 1 1136 5703531 97

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000364.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pcdhb9

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 BC062640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 AY013791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 BC055319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 NM_053134.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 BC062640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 AY013791.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 BC055319.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 NM_053134.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834765 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 51584 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137689 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 BC062640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 AY013791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 BC055319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 NM_053134.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 AK032626.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 BC062640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 AY013791.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 BC055319.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 NM_053134.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 BC062640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 AY013791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 BC055319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 NM_053134.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 BC062640.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 AY013791.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 BC055319.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 NM_053134.3 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628064 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628071 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628069 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628065 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31628088 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08968106 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137689 - -
          Whole Mouse Genome G4122A A_51_P483878 BC062640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 AY013791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 BC055319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 NM_053134.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 AK032626.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 BC062640.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 AY013791.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 BC055319.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 NM_053134.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P483878 AK032626.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P129269 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137689 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13861992 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13861973 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13861984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628120 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31628119 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14037713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628095 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628074 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628064 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13862017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628062 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13861975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628065 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628078 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13862012 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628067 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628088 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31628071 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10455082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10455081 - -
          Affymetrix HT MG-430 PM Array Plate 1422640_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10455080 - -
          Mouse Genome 430 2.0 Array 1422640_PM_at - -
          Mouse Genome 430 2.0 Array 1422640_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93880_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 93880_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800002834.mm.1 TC1800000364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002834.mm.1 NM_053134.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002834.mm.1 NM_053134.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002833.mm.1 TC1800000364.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800002833.mm.1 NM_053134.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800002833.mm.1 NM_053134.3 Refseq
          Mouse Expression Set 430 1422640_PM_at - -
          Mouse Expression Set 430 1422640_at - -
          Mouse Genome 430A 2.0 Array 1422640_at - -
          Mouse Genome 430A 2.0 Array 1422640_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422640_at - -
          Affymetrix HT Mouse Genome 430A Array 1422640_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 714975 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230102 - -
          Illumina mouse-6 v1.0 expression beadchip 4230102 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2711955 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2711955 - -
          mouse-6 v1.1 (Illumina) scl52065.1.27_54-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230102 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52065.1.27_54-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230102 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2711955 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2711955 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl52065.1.27_54-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0