Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00475031_m1
          See other FOXP2 GE Assays ›
          Gene Symbol:
          Foxp2
          Gene Name:
          forkhead box P2
          Gene Aliases:
          2810043D05Rik, AI449000, CAG-16, D0Kist7
          Chromosome Location:
          Chr.6: 14901226 - 15441977 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00475031_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Foxp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001286607.1 NP_001273536.1 7 - 8 1242 73
          NM_053242.4 NP_444472.2 9 - 10 1642 73
          NM_212435.1 NP_997600.1 7 - 8 1305 73
          GenBank mRNA AF339106.1 - 6 - 7 983 73
          AK051668.1 - 9 - 10 1625 73
          AK082361.1 - 9 - 10 1314 73
          AY079003.1 - 9 - 10 1271 73
          BC058960.1 - 7 - 8 1239 6811877 73
          BC062926.1 - 7 - 8 1305 6851153 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600000103.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Foxp2

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          positive regulation of mesenchymal cell proliferation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          skeletal muscle tissue development
          embryo development
          post-embryonic development
          cerebellum development
          lung development
          growth
          vocal learning
          camera-type eye development
          negative regulation of transcription, DNA-templated
          lung alveolus development
          smooth muscle tissue development
          positive regulation of epithelial cell proliferation
          righting reflex
          positive regulation of epithelial cell proliferation involved in lung morphogenesis
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein homodimerization activity
          sequence-specific DNA binding
          metal ion binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 TC0600000103.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 NM_001286607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 NM_053242.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 NM_053242.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 NM_212435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000709.mm.1 NM_212435.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 TC0600000103.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 NM_001286607.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 NM_053242.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 NM_053242.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 NM_212435.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600000710.mm.1 NM_212435.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0