Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00477453_g1
          See other AIRE GE Assays ›
          Gene Symbol:
          Aire
          Gene Name:
          autoimmune regulator (autoimmune polyendocrinopathy candidiasis ectodermal dystrophy)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 78030022 - 78043610 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00477453_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aire
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001271549.1 NP_001258478.1 4 - 5 629 56
          NM_001271550.1 NP_001258479.1 4 - 5 629 56
          NM_001271551.1 NP_001258480.1 4 - 5 629 56
          NM_001271552.1 NP_001258481.1 4 - 5 629 56
          NM_001271553.1 NP_001258482.1 4 - 5 629 56
          NM_001271554.1 NP_001258483.1 4 - 5 629 56
          NM_001271555.1 NP_001258484.1 4 - 5 629 56
          NM_001271556.1 NP_001258485.1 4 - 5 629 56
          NM_001271557.1 NP_001258486.1 4 - 5 629 56
          NM_001271558.1 NP_001258487.1 4 - 5 629 56
          NM_001271559.1 NP_001258488.1 4 - 5 629 56
          NM_009646.2 NP_033776.1 4 - 5 629 56
          NR_073358.1 4 - 5 629 56
          GenBank mRNA AF079536.1 - 4 - 5 599 56
          AF128115.1 - 4 - 5 599 56
          AF128116.1 - 4 - 5 599 56
          AF128117.1 - 4 - 5 599 56
          AF128118.1 - 4 - 5 599 56
          AF128119.1 - 4 - 5 599 56
          AF128120.1 - 4 - 5 599 56
          AF128121.1 - 4 - 5 599 56
          AF128122.1 - 4 - 5 599 56
          AF128123.1 - 4 - 5 599 56
          AF128124.1 - 4 - 5 599 56
          AF128125.1 - 4 - 5 599 56
          AF128772.1 - 4 - 5 542 56
          AF128773.1 - 4 - 5 542 56
          AJ132243.1 - 4 - 5 542 56
          AJ243821.1 - 4 - 5 542 56
          AK153690.1 - 5 - 6 683 56
          BC103510.1 - 4 - 5 591 40039055 56
          BC103511.1 - 4 - 5 591 40039054 56
          BC103518.1 - 4 - 5 591 40039058 56
          EU625343.3 - 4 - 5 559 56
          JF267399.1 - 4 - 5 559 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000002440.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aire

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          humoral immune response
          positive regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          transcription cis-regulatory region binding
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          DNA binding
          chromatin binding
          zinc ion binding
          histone binding
          identical protein binding
          translation regulator activity
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 TC1000002440.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271549.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271550.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271551.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271552.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271553.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271554.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271555.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271556.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271557.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271558.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_001271559.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NM_009646.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1000017743.mm.1 NR_073358.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0