Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00479005_g1
          See other PSMB9 GE Assays ›
          Gene Symbol:
          Psmb9
          Gene Name:
          proteasome (prosome, macropain) subunit, beta type 9 (large multifunctional peptidase 2)
          Gene Aliases:
          Lmp-2, Lmp2
          Chromosome Location:
          Chr.17: 34182030 - 34187330 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00479005_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psmb9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013585.2 NP_038613.1 3 - 4 276 72
          GenBank mRNA AK008429.1 - 3 - 4 267 72
          AK152743.1 - 3 - 4 321 72
          AK171670.1 - 3 - 4 286 72
          BC116364.2 - 3 - 4 276 40100021 72
          CT010227.1 - 3 - 4 255 72
          D44454.1 - 3 - 4 276 72
          D44456.1 - 3 - 4 276 72
          D44457.1 - 3 - 4 276 72
          D44458.1 - 3 - 4 276 72
          D44461.1 - 3 - 4 276 72
          KJ650226.1 - 3 - 4 255 72
          S58203.1 - 3 - 4 276 72
          S59862.1 - 3 - 4 276 72
          U22447.1 - 3 - 4 255 72
          U22448.1 - 3 - 4 255 72
          U22919.1 - 3 - 4 255 72
          U22920.1 - 3 - 4 255 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001902.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psmb9

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          proteolysis
          proteasomal ubiquitin-independent protein catabolic process
          antigen processing and presentation
          proteasome-mediated ubiquitin-dependent protein catabolic process
          proteolysis involved in cellular protein catabolic process
          endopeptidase activity
          threonine-type endopeptidase activity
          peptidase activity
          hydrolase activity
          proteasome binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152611 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152609 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 U22920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 D44461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 D44457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 S58203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 U22920.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 D44461.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 D44457.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 S58203.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2038983 BC032210.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 344038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 441539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161174 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125409 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 U22920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 D44461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 D44457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 S58203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 S59862.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 U22920.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 D44461.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 D44457.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 S58203.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 U22920.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 D44461.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 D44457.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 S58203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 U22920.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 D44461.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 D44457.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 S58203.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152587 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152582 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152583 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152581 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152598 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152590 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152580 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152600 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152572 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152584 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152579 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31152576 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125409 - -
          Whole Mouse Genome G4122A A_51_P369803 U22920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 D44461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 D44457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 S58203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 S59862.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 U22920.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 D44461.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 D44457.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 S58203.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369803 S59862.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132642 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125409 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13515780 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13515759 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13515758 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13515750 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152610 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152609 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31152611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31152576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13344255 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13515761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152600 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152607 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152605 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13344256 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13240028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152562 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13387804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152559 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13098023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152567 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31152592 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450150 - -
          Affymetrix HT MG-430 PM Array Plate 1450696_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450145 - -
          Mouse Genome 430 2.0 Array 1450696_PM_at - -
          Mouse Genome 430 2.0 Array 1450696_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16912_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16912_at - -
          Affymetrix Mouse Clariom S Array PSR1700017770.mm.1 TC1700001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017770.mm.1 NM_013585.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017770.mm.1 NM_013585.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017772.mm.1 TC1700001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017772.mm.1 NM_013585.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017772.mm.1 NM_013585.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017776.mm.1 TC1700001902.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700017776.mm.1 NM_013585.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700017776.mm.1 NM_013585.2 Refseq
          Mouse Expression Set 430 1450696_at - -
          Mouse Expression Set 430 1450696_PM_at - -
          Murine Genome U74v2 Set 93085_at - -
          Murine Genome U74v2 Set 93085_PM_at - -
          Mouse Genome 430A 2.0 Array 1450696_at - -
          Mouse Genome 430A 2.0 Array 1450696_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450696_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450696_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6980471 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006980471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1242466 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1242466 - -
          mouse-6 v1.1 (Illumina) scl50027.4.1_35-S - -
          Illumina Sentrix mouse-6.v1 bead array 6980471 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50027.4.1_35-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6980471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1242466 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1242466 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50027.4.1_35-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-x8sd2:80/100.66.134.75:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0