Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480133_m1
          See other PRMT1 GE Assays ›
          Gene Symbol:
          Prmt1
          Gene Name:
          protein arginine N-methyltransferase 1
          Gene Aliases:
          6720434D09Rik, AW214366, Hrmt1l2, Mrmt1
          Chromosome Location:
          Chr.7: 44976755 - 44986420 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480133_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prmt1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019830.3 NP_062804.1 1 - 2 138 59
          NR_045521.1 1 - 2 138 59
          GenBank mRNA AF232716.1 - 1 - 2 50 59
          BC051953.1 - 1 - 2 74 5692474 59
          BC062964.1 - 1 - 2 74 5692474 59
          BU524421.1 - 1 - 2 138 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002980.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC051953.1, BC062964.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prmt1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of transcription, DNA-templated
          protein methylation
          histone methylation
          peptidyl-arginine methylation
          peptidyl-arginine methylation, to asymmetrical-dimethyl arginine
          neuron projection development
          methylation
          peptidyl-arginine N-methylation
          peptidyl-arginine omega-N-methylation
          histone H4-R3 methylation
          negative regulation of megakaryocyte differentiation
          rRNA (adenine-N6,N6-)-dimethyltransferase activity
          RNA binding
          protein C-terminal carboxyl O-methyltransferase activity
          tRNA (guanine-N2-)-methyltransferase activity
          protein binding
          methyltransferase activity
          C-methyltransferase activity
          N-methyltransferase activity
          O-methyltransferase activity
          S-methyltransferase activity
          RNA methyltransferase activity
          mRNA methyltransferase activity
          tRNA methyltransferase activity
          protein methyltransferase activity
          2-polyprenyl-6-methoxy-1,4-benzoquinone methyltransferase activity
          histone-arginine N-methyltransferase activity
          rRNA methyltransferase activity
          rRNA (uridine-2'-O-)-methyltransferase activity
          rRNA (adenine-N6-)-methyltransferase activity
          DNA-methyltransferase activity
          tRNA (guanine-N1-)-methyltransferase activity
          rRNA (cytosine-C5-)-methyltransferase activity
          selenocysteine methyltransferase activity
          arginine N-methyltransferase activity
          protein-arginine N-methyltransferase activity
          [cytochrome c]-arginine N-methyltransferase activity
          lysine N-methyltransferase activity
          protein-lysine N-methyltransferase activity
          tRNA (uracil) methyltransferase activity
          tRNA (guanine) methyltransferase activity
          tRNA (adenine) methyltransferase activity
          tRNA (cytosine) methyltransferase activity
          tRNA (cytosine-5-)-methyltransferase activity
          tRNA (adenine-N1-)-methyltransferase activity
          rRNA (adenine) methyltransferase activity
          rRNA (cytosine) methyltransferase activity
          rRNA (guanine) methyltransferase activity
          rRNA (uridine) methyltransferase activity
          transferase activity
          protein C-terminal leucine carboxyl O-methyltransferase activity
          1-phenanthrol methyltransferase activity
          protein-arginine N5-methyltransferase activity
          snoRNP binding
          methylarsonite methyltransferase activity
          dimethylarsinite methyltransferase activity
          4,5-dihydroxybenzo(a)pyrene methyltransferase activity
          1-hydroxypyrene methyltransferase activity
          1-hydroxy-6-methoxypyrene methyltransferase activity
          protein-arginine omega-N monomethyltransferase activity
          protein-arginine omega-N asymmetric methyltransferase activity
          histone methyltransferase activity
          identical protein binding
          demethylmenaquinone methyltransferase activity
          cobalt-precorrin-6B C5-methyltransferase activity
          cobalt-precorrin-7 C15-methyltransferase activity
          cobalt-precorrin-5B C1-methyltransferase activity
          cobalt-precorrin-3 C17-methyltransferase activity
          dimethylamine methyltransferase activity
          hydroxyneurosporene-O-methyltransferase activity
          tRNA (adenine-57, 58-N(1)-) methyltransferase activity
          methylamine-specific methylcobalamin:coenzyme M methyltransferase activity
          trimethylamine methyltransferase activity
          methanol-specific methylcobalamin:coenzyme M methyltransferase activity
          monomethylamine methyltransferase activity
          histone methyltransferase activity (H4-R3 specific)
          P-methyltransferase activity
          Se-methyltransferase activity
          2-phytyl-1,4-naphthoquinone methyltransferase activity
          tRNA (uracil-2'-O-)-methyltransferase activity
          tRNA (cytosine-2'-O-)-methyltransferase activity
          phosphomethylethanolamine N-methyltransferase activity
          tRNA (cytosine-3-)-methyltransferase activity
          rRNA (cytosine-2'-O-)-methyltransferase activity
          rRNA (cytosine-N4-)-methyltransferase activity
          trihydroxyferuloyl spermidine O-methyltransferase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026728.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026726.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026726.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026726.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026726.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026726.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026727.mm.1 NR_045521.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 TC0700002980.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_001252476.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_001252476.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_001252477.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_001252477.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_019830.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NM_019830.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NR_045521.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700026729.mm.1 NR_045521.1 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 658449 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0