Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00480200_m1
          See other CLCF1 GE Assays ›
          Gene Symbol:
          Clcf1
          Gene Name:
          cardiotrophin-like cytokine factor 1
          Gene Aliases:
          Bsf3, CLC
          Chromosome Location:
          Chr.19: 4214392 - 4222615 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00480200_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Clcf1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019952.3 NP_064336.1 1 - 2 111 107
          GenBank mRNA AF176913.1 - 1 - 2 112 107
          AK137396.1 - 239 107
          AK166421.1 - 115 107
          AK170969.1 - 1 - 2 129 107
          BC104258.1 - 1 - 2 86 40039794 107
          BC104259.1 - 1 - 2 86 40039795 107

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900000035.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AF176913.1, AK170969.1, BC104258.1, BC104259.1, NM_019952.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Clcf1

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of immunoglobulin production
          positive regulation of type 2 immune response
          cell surface receptor signaling pathway
          receptor signaling pathway via JAK-STAT
          positive regulation of cell population proliferation
          B cell differentiation
          positive regulation of B cell proliferation
          positive regulation of tyrosine phosphorylation of STAT protein
          negative regulation of neuron apoptotic process
          positive regulation of isotype switching to IgE isotypes
          positive regulation of astrocyte differentiation
          regulation of inflammatory response
          positive regulation of corticotropin-releasing hormone secretion
          regulation of protein kinase B signaling
          regulation of ERK1 and ERK2 cascade
          negative regulation of motor neuron apoptotic process
          signaling receptor binding
          cytokine activity
          ciliary neurotrophic factor receptor binding
          protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1900000494.mm.1 TC1900000035.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000494.mm.1 NM_019952.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000495.mm.1 TC1900000035.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000495.mm.1 NM_019952.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1900000498.mm.1 TC1900000035.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900000498.mm.1 NM_019952.5 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0