Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114085.1 | NP_001107557.1 | 6 - 7 | 842 | 87 | |
NM_001285503.1 | NP_001272432.1 | 6 - 7 | 842 | 87 | ||
NM_001285504.1 | NP_001272433.1 | 5 - 6 | 693 | 87 | ||
NM_023317.2 | NP_075806.2 | 6 - 7 | 842 | 87 | ||
GenBank mRNA | AF290473.1 | - | 4 - 5 | 576 | 87 | |
AF322073.1 | - | 5 - 6 | 749 | 87 | ||
AK012830.1 | - | 6 - 7 | 772 | 87 | ||
AK012861.1 | - | 6 - 7 | 790 | 87 | ||
AK037289.1 | - | 6 - 7 | 785 | 87 | ||
AK146982.1 | - | 6 - 7 | 772 | 87 | ||
AK150826.1 | - | 6 - 7 | 832 | 87 | ||
AK166858.1 | - | 5 - 6 | 672 | 87 | ||
AK169743.1 | - | 6 - 7 | 772 | 87 | ||
BC023267.1 | - | 6 - 7 | 821 | 3672565 | 87 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |