Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481057_m1
          See other MRPL46 GE Assays ›
          Gene Symbol:
          Mrpl46
          Gene Name:
          mitochondrial ribosomal protein L46
          Gene Aliases:
          3110052F15Rik, C15orf4, C80107, LIECG2, P2ECSL
          Chromosome Location:
          Chr.7: 78775341 - 78783089 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481057_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mrpl46
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_023331.2 NP_075820.1 3 - 4 641 77
          GenBank mRNA AF217090.1 - 3 - 4 608 77
          AK006415.1 - 2 - 3 297 77
          AK014205.1 - 3 - 4 618 77
          BC061094.1 - 3 - 4 641 6741290 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700003547.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mrpl46

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          structural constituent of ribosome
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257247 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257249 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126320 BC061094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126320 AF217090.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126320 AK006415.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126320 NM_023331.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P126320 AK014205.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 709076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 547290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 461909 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 648199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836309 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 393207 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71585 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P124849 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126320 BC061094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126320 AF217090.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126320 AK006415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126320 NM_023331.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P126320 AK014205.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257227 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257225 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257230 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257235 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25257241 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06252042 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P124849 - -
          Whole Mouse Genome G4122A A_51_P335916 BC061094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335916 AF217090.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335916 NM_023331.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P335916 AK014205.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116814 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126320 BC061094.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126320 AF217090.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126320 AK006415.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126320 NM_023331.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126320 AK014205.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P124849 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957637 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05957639 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06229308 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257246 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257249 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25257247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25257170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257138 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06236689 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473817 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257232 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473820 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05994242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473812 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473823 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06356815 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257218 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257180 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05994233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05994248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25257235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06473833 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10564697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564695 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564694 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564693 - -
          Affymetrix Mouse Gene 1.0 ST Array 10564696 - -
          Affymetrix HT MG-430 PM Array Plate 1449004_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10564692 - -
          Mouse Genome 430 2.0 Array 1449004_at - -
          Mouse Genome 430 2.0 Array 1449004_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67308_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67308_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700029951.mm.1 TC0700003547.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029951.mm.1 NM_023331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029951.mm.1 NM_023331.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029953.mm.1 TC0700003547.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029953.mm.1 NM_023331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029953.mm.1 NM_023331.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029955.mm.1 TC0700003547.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029955.mm.1 NM_023331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029955.mm.1 NM_023331.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029952.mm.1 TC0700003547.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029952.mm.1 NM_023331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029952.mm.1 NM_023331.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029954.mm.1 TC0700003547.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700029954.mm.1 NM_023331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700029954.mm.1 NM_023331.2 Refseq
          Mouse Expression Set 430 1449004_at - -
          Mouse Expression Set 430 1449004_PM_at - -
          Murine Genome U74v2 Set 104567_PM_at - -
          Murine Genome U74v2 Set 162436_at - -
          Murine Genome U74v2 Set 162436_PM_at - -
          Murine Genome U74v2 Set 104567_at - -
          Murine Genome U74v2 Set 168053_PM_f_at - -
          Murine Genome U74v2 Set 168053_f_at - -
          Mouse Genome 430A 2.0 Array 1449004_PM_at - -
          Mouse Genome 430A 2.0 Array 1449004_at - -
          Affymetrix HT Mouse Genome 430A Array 1449004_at - -
          Affymetrix HT Mouse Genome 430A Array 1449004_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 580416 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 360736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000360736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740922 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740922 - -
          mouse-6 v1.1 (Illumina) scl31161.4.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 360736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31161.4.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 360736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740922 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740922 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl31161.4.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0