Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00481252_m1
          See other AAED1 GE Assays ›
          Gene Symbol:
          Aaed1
          Gene Name:
          AhpC/TSA antioxidant enzyme domain containing 1
          Gene Aliases:
          1110018J18Rik, AI849003
          Chromosome Location:
          Chr.13: 64291836 - 64312710 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00481252_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aaed1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025370.2 NP_079646.1 2 - 3 285 95
          GenBank mRNA AK003782.1 - 2 - 3 286 95
          AK006589.1 - 2 - 3 140 95
          AK145073.1 - 283 95
          AK145867.1 - 286 95
          AK148379.1 - 286 95
          AK168701.1 - 164 95

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002201.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aaed1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098293 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009351 AK168581.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009351 AK168701.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009351 AK145073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009351 AK145867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2009351 AK006589.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149867 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 29301 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 953097 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 375509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 682350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 427238 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 720982 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852476 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 19595 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 903760 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 171015 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 616182 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 98239 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151069 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 45658 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117909 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P129238 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009351 AK168581.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009351 AK168701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009351 AK145073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009351 AK145867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2009351 AK006589.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016193 AK168701.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2016193 AK145867.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098310 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29098295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P129238 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07900882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02709655 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P117909 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02709656 - -
          Whole Mouse Genome G4122A A_51_P361049 AK148379.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361049 AK168581.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361049 AK168701.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361049 AK145073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P361049 AK145867.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108423 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02709655 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P129238 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P117909 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10929178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10929174 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10875919 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098291 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29098292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098269 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29098304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10898392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098206 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098213 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098288 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10882289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098296 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098230 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098304 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098326 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11057252 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098273 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098269 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098229 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098238 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10929178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10455014 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098266 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11000739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10939497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11093979 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098306 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098299 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098300 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098210 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098310 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10498793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10839329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10682507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11083158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098226 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11063275 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10690842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098207 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10682506 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10929183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29098267 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10410123 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410122 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410118 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410117 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410121 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410119 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410120 - -
          Affymetrix Mouse Gene 1.0 ST Array 10410116 - -
          Affymetrix HT MG-430 PM Array Plate 1421018_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434441_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10410115 - -
          Mouse Genome 430 2.0 Array 1434441_PM_at - -
          Mouse Genome 430 2.0 Array 1434441_at - -
          Mouse Genome 430 2.0 Array 1421018_PM_at - -
          Mouse Genome 430 2.0 Array 1421018_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66129_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66129_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300014218.mm.1 TC1300002201.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300014218.mm.1 NM_025370.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300014218.mm.1 NM_025370.2 Refseq
          Mouse Expression Set 430 1421018_PM_at - -
          Mouse Expression Set 430 1421018_at - -
          Mouse Expression Set 430 1434441_PM_at - -
          Mouse Expression Set 430 1434441_at - -
          Mouse Genome 430A 2.0 Array 1421018_at - -
          Mouse Genome 430A 2.0 Array 1421018_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434441_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434441_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 685724 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 4810671 - -
          Illumina mouse-6 v1.0 expression beadchip 70161 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004810671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000070161 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2647669 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2674655 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2647669 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2674655 - -
          mouse-6 v1.1 (Illumina) scl0003641.1_23-S - -
          mouse-6 v1.1 (Illumina) scl066129.2_5-S - -
          Illumina Sentrix mouse-6.v1 bead array 70161 - -
          Illumina Sentrix mouse-6.v1 bead array 4810671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003641.1_23-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl066129.2_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4810671 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 70161 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2674655 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2647669 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2647669 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2674655 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0