Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00482209_m1
          See other NUDT16L1 GE Assays ›
          Gene Symbol:
          Nudt16l1
          Gene Name:
          nudix (nucleoside diphosphate linked moiety X)-type motif 16-like 1
          Gene Aliases:
          1110001K21Rik, 5330437I08Rik, Sdos
          Chromosome Location:
          Chr.16: 4939111 - 4941021 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00482209_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nudt16l1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025839.3 NP_080115.1 2 - 3 441 85
          GenBank mRNA AF435792.1 - 2 - 3 453 85
          AK002378.1 - 2 - 3 434 85
          AK003241.1 - 2 - 3 442 85
          AK007897.1 - 2 - 3 437 85
          AK159841.1 - 2 - 3 434 85
          AK186853.1 - 1 - 2 151 85
          BC028494.1 - 2 - 3 434 1511165 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1600000040.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nudt16l1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          RNA binding
          hydrolase activity
          snoRNA binding
          m7G(5')pppN diphosphatase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512784 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512787 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512788 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 BC025569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK003241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK007897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK019936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK159841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 BC025569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK003241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK007897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK019936.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P369516 AK159841.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112479 BC025569.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112479 BC028494.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112479 AK003241.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112479 AK007897.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2112479 AK019936.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 600395 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P160416 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112479 BC025569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112479 BC028494.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112479 AK003241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112479 AK007897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2112479 AK019936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 BC025569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK003241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK007897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK019936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK159841.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 BC025569.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK003241.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK007897.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK019936.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P369516 AK159841.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512792 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512789 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512780 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512793 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512795 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512791 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512797 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512786 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30512777 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P160416 - -
          Whole Mouse Genome G4122A A_51_P369516 BC025569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK003241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK007897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK019936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK159841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 BC025569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK003241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK007897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK019936.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P369516 AK159841.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434570 BC025569.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434570 BC028494.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434570 AK003241.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434570 AK007897.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434570 AK159841.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140149 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P160416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12824966 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12824949 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12824956 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12825032 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12824998 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512785 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512787 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512789 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512788 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512786 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30512784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512800 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30512778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12824977 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12824949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12824971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30512784 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10433336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433332 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433334 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10433339 - -
          Affymetrix HT MG-430 PM Array Plate 1428049_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1437624_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10433331 - -
          Mouse Genome 430 2.0 Array 1437624_PM_x_at - -
          Mouse Genome 430 2.0 Array 1437624_x_at - -
          Mouse Genome 430 2.0 Array 1428049_PM_a_at - -
          Mouse Genome 430 2.0 Array 1428049_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66911_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66911_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1600000357.mm.1 TC1600000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000357.mm.1 NM_025839.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000358.mm.1 TC1600000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000358.mm.1 NM_025839.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000359.mm.1 TC1600000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000359.mm.1 NM_025839.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000355.mm.1 TC1600000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000355.mm.1 NM_025839.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1600000356.mm.1 TC1600000040.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1600000356.mm.1 NM_025839.4 Refseq
          Mouse Expression Set 430 1437624_PM_x_at - -
          Mouse Expression Set 430 1428049_a_at - -
          Mouse Expression Set 430 1437624_x_at - -
          Mouse Expression Set 430 1428049_PM_a_at - -
          Murine Genome U74v2 Set 98061_at - -
          Murine Genome U74v2 Set 98061_PM_at - -
          Mouse Genome 430A 2.0 Array 1428049_a_at - -
          Mouse Genome 430A 2.0 Array 1437624_x_at - -
          Mouse Genome 430A 2.0 Array 1437624_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1428049_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437624_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1428049_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437624_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1428049_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2360438 - -
          Illumina mouse-6 v1.0 expression beadchip 2360438 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002360438 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2767227 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2767227 - -
          mouse-6 v1.1 (Illumina) scl49474.3_50-S - -
          Illumina Sentrix mouse-6.v1 bead array 2360438 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl49474.3_50-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2360438 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2767227 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2767227 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl49474.3_50-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-784b477dbd-pq4vn:80/100.66.130.62:80.
          git-commit: d41a97e986437cfbf980934f14197a4b4d3eb741
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0