Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkain1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025998.3 | NP_080274.2 | 4 - 5 | 627 | 79 | |
GenBank mRNA | AK011164.1 | - | 4 - 5 | 760 | 79 | |
AK011850.1 | - | 4 - 5 | 558 | 79 | ||
AK076029.1 | - | 1 - 2 | 115 | 79 | ||
AK184838.1 | - | 1 - 2 | 80 | 79 | ||
AK187454.1 | - | 1 - 2 | 80 | 79 | ||
AK205247.1 | - | 1 - 2 | 80 | 79 | ||
AK205326.1 | - | 1 - 2 | 80 | 79 | ||
AK206217.1 | - | 1 - 2 | 80 | 79 | ||
AK206243.1 | - | 1 - 2 | 80 | 79 | ||
AK208705.1 | - | 1 - 2 | 80 | 79 | ||
AK208715.1 | - | 1 - 2 | 80 | 79 | ||
AK209134.1 | - | 1 - 2 | 80 | 79 | ||
AK209985.1 | - | 1 - 2 | 80 | 79 | ||
AK210264.1 | - | 1 - 2 | 80 | 79 | ||
AK211305.1 | - | 1 - 2 | 80 | 79 | ||
AK211393.1 | - | 1 - 2 | 80 | 79 | ||
AK211541.1 | - | 1 - 2 | 80 | 79 | ||
AK213620.1 | - | 1 - 2 | 80 | 79 | ||
AK214155.1 | - | 1 - 2 | 80 | 79 | ||
AK215330.1 | - | 1 - 2 | 80 | 79 | ||
AK216110.1 | - | 1 - 2 | 80 | 79 | ||
AK216558.1 | - | 1 - 2 | 80 | 79 | ||
AK216625.1 | - | 1 - 2 | 80 | 79 | ||
AK216635.1 | - | 1 - 2 | 80 | 79 | ||
AK216972.1 | - | 1 - 2 | 80 | 79 | ||
AK217347.1 | - | 1 - 2 | 80 | 79 | ||
AK218646.1 | - | 1 - 2 | 80 | 79 | ||
AK218997.1 | - | 1 - 2 | 80 | 79 | ||
BC116391.1 | - | 4 - 5 | 557 | 40101865 | 79 | |
BC116392.1 | - | 4 - 5 | 574 | 79 | ||
EF058048.2 | - | 4 - 5 | 680 | 79 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |