Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485418_m1
          See other MYT1L GE Assays ›
          Gene Symbol:
          Myt1l
          Gene Name:
          myelin transcription factor 1-like
          Gene Aliases:
          2900046C06Rik, 2900093J19Rik, C630034G21Rik, Nztf1, Pmng1, Png-1, mKIAA1106
          Chromosome Location:
          Chr.12: 29528311 - 29923216 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485418_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Myt1l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001093775.1 NP_001087244.1 21 - 22 3992 63
          NM_001093776.1 NP_001087245.1 21 - 22 3898 63
          NM_001093778.1 NP_001087247.1 21 - 22 3983 63
          NM_008666.3 NP_032692.2 21 - 22 3986 63
          GenBank mRNA AB093283.1 - 18 - 19 3345 63
          AB212898.1 - 20 - 21 3590 63
          AF004295.1 - 21 - 22 3802 63
          BC079651.1 - 21 - 22 3867 63
          BC094438.1 - 17 - 18 3223 30692780 63
          BC131677.1 - 20 - 21 3587 40111036 63
          U86338.1 - 21 - 22 3714 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1200000247.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Myt1l

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription by RNA polymerase II
          multicellular organism development
          nervous system development
          cell differentiation
          positive regulation of transcription by RNA polymerase II
          DNA binding
          DNA-binding transcription factor activity
          transcription coactivator activity
          protein binding
          zinc ion binding
          retinoic acid-responsive element binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 TC1200000247.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093775.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093775.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093776.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093776.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093778.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_001093778.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_008666.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1200001904.mm.1 NM_008666.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0