Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00485667_m1
          See other RELB GE Assays ›
          Gene Symbol:
          Relb
          Gene Name:
          avian reticuloendotheliosis viral (v-rel) oncogene related B
          Gene Aliases:
          shep
          Chromosome Location:
          Chr.7: 19606217 - 19629438 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00485667_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Relb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290457.1 NP_001277386.1 5 - 6 882 66
          NM_009046.2 NP_033072.2 6 - 7 891 66
          GenBank mRNA AK146932.1 - 6 - 7 892 66
          BC019765.1 - 6 - 7 891 3584200 66
          BC034523.1 - 5 - 6 695 4485540 66
          BC117792.2 - 6 - 7 66
          BC117793.1 - 6 - 7 827 40092162 66
          DQ020178.1 - 5 - 6 543 66
          DQ020179.1 - 5 - 6 1659 66
          DQ020180.1 - 7 - 8 1699 66
          DQ020181.1 - 6 - 7 1725 66
          M83380.1 - 6 - 7 728 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002491.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Relb

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          inflammatory response
          I-kappaB kinase/NF-kappaB signaling
          positive regulation of gene expression
          antigen processing and presentation
          negative regulation of interferon-beta production
          circadian regulation of gene expression
          response to cytokine
          NIK/NF-kappaB signaling
          T-helper 1 type immune response
          myeloid dendritic cell differentiation
          T-helper 1 cell differentiation
          innate immune response
          negative regulation of transcription, DNA-templated
          positive regulation of transcription by RNA polymerase II
          rhythmic process
          cellular response to osmotic stress
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          DNA binding
          chromatin binding
          DNA-binding transcription factor activity
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700021959.mm.1 TC0700002491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021959.mm.1 NM_009046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021959.mm.1 NM_009046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021958.mm.1 TC0700002491.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021958.mm.1 NM_009046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021958.mm.1 NM_009046.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0