Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00486655_m1
          See other CALM1 GE Assays ›
          Gene Symbol:
          Calm1
          Gene Name:
          calmodulin 1
          Gene Aliases:
          AI256814, AI327027, AI461935, AL024000, CaM, Calm
          Chromosome Location:
          Chr.12: 100199541 - 100209806 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00486655_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Calm1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009790.4 NP_033920.1 2 - 3 204 111
          GenBank mRNA AK004673.1 - 2 - 3 218 111
          AK013695.1 - 2 - 3 213 111
          AK088141.1 - 2 - 3 214 111
          AK150288.1 - 2 - 3 214 111
          AK150978.1 - 2 - 3 219 111
          AK151001.1 - 2 - 3 214 111
          AK151552.1 - 2 - 3 214 111
          AK151784.1 - 2 - 3 219 111
          AK151923.1 - 2 - 3 217 111
          AK151992.1 - 2 - 3 219 111
          AK152148.1 - 2 - 3 217 111
          AK152715.1 - 2 - 3 213 111
          AK152719.1 - 2 - 3 214 111
          AK152850.1 - 2 - 3 217 111
          AK152897.1 - 2 - 3 214 111
          AK153004.1 - 2 - 3 213 111
          AK153348.1 - 2 - 3 217 111
          AK153426.1 - 2 - 3 217 111
          AK153546.1 - 2 - 3 217 111
          AK159762.1 - 2 - 3 220 111
          AK160057.1 - 2 - 3 220 111
          AK160508.1 - 2 - 3 219 111
          AK161302.1 - 2 - 3 175 111
          AK162314.1 - 2 - 3 221 111
          AK166308.1 - 2 - 3 214 111
          AK167353.1 - 2 - 3 219 111
          AK168002.1 - 2 - 3 217 111
          AK168241.1 - 2 - 3 173 111
          AK168663.1 - 2 - 3 213 111
          AK168803.1 - 2 - 3 217 111
          AK169027.1 - 2 - 3 219 111
          AK169055.1 - 2 - 3 217 111
          AK169640.1 - 2 - 3 219 111
          BC054805.1 - 2 - 3 204 6838207 111
          X61432.1 - 2 - 3 57 111

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK004673.1, AK013695.1, AK088141.1, AK150288.1, AK150978.1, AK151001.1, AK151552.1, AK151784.1, AK151923.1, AK151992.1, AK152148.1, AK152715.1, AK152719.1, AK152850.1, AK152897.1, AK153004.1, AK153348.1, AK153426.1, AK153546.1, AK159762.1, AK160057.1, AK160508.1, AK161302.1, AK162314.1, AK166308.1, AK167353.1, AK168002.1, AK168241.1, AK168663.1, AK168803.1, AK169027.1, AK169055.1, AK169640.1, BC054805.1, NM_009790.4, X61432.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Calm1

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          positive regulation of DNA binding
          regulation of catalytic activity
          calcium ion binding
          adenylate cyclase binding
          protein domain specific binding
          enzyme regulator activity
          nitric-oxide synthase regulator activity
          type 3 metabotropic glutamate receptor binding
          phosphatidylinositol 3-kinase binding
          transmembrane transporter binding
          protein N-terminus binding
          calcium-dependent protein binding
          nitric-oxide synthase binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1200006933.mm.1 TC1200000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006933.mm.1 NM_009790.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006931.mm.1 TC1200000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006931.mm.1 NM_009790.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006937.mm.1 TC1200000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006937.mm.1 NM_009790.5 Refseq
          Affymetrix Mouse Clariom S Array PSR1200006934.mm.1 TC1200000994.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1200006934.mm.1 NM_009790.5 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 692243 - -
          ABI Mouse Genome Survey Microarray 910929 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0