Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00487418_m1
          See other FKBP8 GE Assays ›
          Gene Symbol:
          Fkbp8
          Gene Name:
          FK506 binding protein 8
          Gene Aliases:
          38kDa, FKBP-38, FKBP-8, Fkbp38
          Chromosome Location:
          Chr.8: 70527743 - 70535328 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00487418_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fkbp8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111066.1 NP_001104536.1 2 - 3 380 96
          NM_001199631.1 NP_001186560.1 2 - 3 476 96
          NM_010223.2 NP_034353.2 2 - 3 380 96
          GenBank mRNA AB154247.1 - 1 - 2 122 96
          AF030635.1 - 1 - 2 150 96
          AK088739.1 - 2 - 3 315 96
          AK146122.1 - 2 - 3 375 96
          AK152198.1 - 2 - 3 376 96
          AK153000.1 - 2 - 3 373 96
          AK158155.1 - 2 - 3 363 96
          AK165281.1 - 2 - 3 373 96
          AK167663.1 - 2 - 3 363 96
          AK216136.1 - 1 - 2 190 96
          AY225340.1 - 1 - 2 263 96
          AY278608.1 - 2 - 3 339 96
          BC003739.1 - 1 - 2 296 3601511 96
          BC027808.1 - 2 - 3 330 5324380 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800000753.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fkbp8

          Gene Ontology Categories:

          Function(s) Process(es)

          protein peptidyl-prolyl isomerization
          cell fate specification
          protein folding
          apoptotic process
          smoothened signaling pathway
          dorsal/ventral pattern formation
          regulation of gene expression
          dorsal/ventral neural tube patterning
          neural tube development
          regulation of BMP signaling pathway
          positive regulation of BMP signaling pathway
          multicellular organism growth
          camera-type eye development
          negative regulation of apoptotic process
          chaperone-mediated protein folding
          peptidyl-prolyl cis-trans isomerase activity
          FK506 binding
          isomerase activity
          identical protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 TC0800000753.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_001111066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_001111066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_001199631.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_001199631.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_010223.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005318.mm.1 NM_010223.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005310.mm.1 TC0800000753.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005310.mm.1 NM_001111066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005310.mm.1 NM_001111066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005310.mm.1 NM_010223.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005310.mm.1 NM_010223.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005311.mm.1 TC0800000753.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005311.mm.1 NM_001111066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005311.mm.1 NM_001111066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005311.mm.1 NM_010223.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005311.mm.1 NM_010223.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 TC0800000753.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_001111066.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_001111066.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_001199631.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_001199631.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_010223.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800005316.mm.1 NM_010223.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 402075 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-f85dfd666-px2hc:80/100.66.130.62:80.
          git-commit: 2ca7b3ab9a2195be9b417413331e4e963d93e17c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0