Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00488871_m1
          See other SPO11 GE Assays ›
          Gene Symbol:
          Spo11
          Gene Name:
          SPO11 meiotic protein covalently bound to DSB homolog (S. cerevisiae)
          Gene Aliases:
          AI449549
          Chromosome Location:
          Chr.2: 172977222 - 172993576 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00488871_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spo11
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001083959.1 NP_001077428.1 2 - 3 276 128
          NM_001083960.1 NP_001077429.1 2 - 3 276 128
          NM_012046.2 NP_036176.1 3 - 4 390 128
          GenBank mRNA AF126400.1 - 2 - 3 276 128
          AF149309.1 - 2 - 3 239 128
          AF163053.1 - 2 - 3 276 128
          AF163054.1 - 2 - 3 276 128
          AF165313.1 - 2 - 3 269 128
          AF167439.1 - 2 - 3 269 128
          AF169386.1 - 3 - 4 387 128
          AF173848.1 - 3 - 4 383 128
          AK088498.1 - 2 - 3 593 128

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002675.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Spo11

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA catabolic process, endonucleolytic
          ovarian follicle development
          DNA metabolic process
          homologous chromosome pairing at meiosis
          synaptonemal complex assembly
          reciprocal meiotic recombination
          male meiosis I
          spermatid development
          protein localization to chromosome
          oogenesis
          meiotic cell cycle
          DNA binding
          catalytic activity
          ATP binding
          hydrolase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 TC0200002675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_001083959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_001083959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_001083960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_001083960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_012046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021764.mm.1 NM_012046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 TC0200002675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_001083959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_001083959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_001083960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_001083960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_012046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021763.mm.1 NM_012046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 TC0200002675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_001083959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_001083959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_001083960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_001083960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_012046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021767.mm.1 NM_012046.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021769.mm.1 TC0200002675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021769.mm.1 NM_001083959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021769.mm.1 NM_001083959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 TC0200002675.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_001083959.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_001083959.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_001083960.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_001083960.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_012046.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021770.mm.1 NM_012046.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b7b59cd5b-kqhbv:80/100.66.135.122:80.
          git-commit: 87deb889a5bb48f859ebd9970e87be562fd9396c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0