Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00489638_m1
          See other SMAD3 GE Assays ›
          Gene Symbol:
          Smad3
          Gene Name:
          SMAD family member 3
          Gene Aliases:
          AU022421, Madh3
          Chromosome Location:
          Chr.9: 63646766 - 63757994 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00489638_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Smad3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016769.4 NP_058049.3 5 - 6 973 71
          GenBank mRNA AB008192.1 - 5 - 6 682 71
          AF016189.1 - 5 - 6 949 71
          AK041743.1 - 5 - 6 494 71
          AK048626.1 - 5 - 6 973 71
          AK083158.1 - 5 - 6 581 71
          BC066850.1 - 5 - 6 659 30432720 71

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900002480.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Smad3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          skeletal system development
          osteoblast differentiation
          response to hypoxia
          in utero embryonic development
          mesoderm formation
          somitogenesis
          liver development
          negative regulation of protein phosphorylation
          heart looping
          osteoblast development
          immune system development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          transport
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          immune response
          transforming growth factor beta receptor signaling pathway
          SMAD protein complex assembly
          gastrulation
          endoderm development
          negative regulation of cell population proliferation
          embryonic pattern specification
          positive regulation of gene expression
          positive regulation of alkaline phosphatase activity
          positive regulation of epithelial to mesenchymal transition
          regulation of striated muscle tissue development
          regulation of transforming growth factor beta receptor signaling pathway
          mitigation of host defenses by virus
          signal transduction involved in regulation of gene expression
          negative regulation of cell growth
          positive regulation of cell migration
          positive regulation of bone mineralization
          thyroid gland development
          positive regulation of chondrocyte differentiation
          positive regulation of interleukin-1 beta production
          regulation of transforming growth factor beta2 production
          positive regulation of transforming growth factor beta3 production
          activin receptor signaling pathway
          negative regulation of osteoblast proliferation
          intracellular signal transduction
          nodal signaling pathway
          T cell activation
          negative regulation of protein catabolic process
          negative regulation of apoptotic process
          cell-cell junction organization
          negative regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of transcription by RNA polymerase II
          paraxial mesoderm morphogenesis
          developmental growth
          embryonic foregut morphogenesis
          embryonic cranial skeleton morphogenesis
          regulation of epithelial cell proliferation
          negative regulation of inflammatory response
          regulation of immune response
          protein stabilization
          positive regulation of positive chemotaxis
          regulation of binding
          positive regulation of stress fiber assembly
          positive regulation of focal adhesion assembly
          pericardium development
          transdifferentiation
          SMAD protein signal transduction
          negative regulation of wound healing
          lens fiber cell differentiation
          positive regulation of canonical Wnt signaling pathway
          extrinsic apoptotic signaling pathway
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of extracellular matrix assembly
          transcription cis-regulatory region binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          cis-regulatory region sequence-specific DNA binding
          DNA binding
          chromatin binding
          double-stranded DNA binding
          DNA-binding transcription factor activity
          transforming growth factor beta receptor binding
          protein binding
          collagen binding
          beta-catenin binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein kinase binding
          phosphatase binding
          chromatin DNA binding
          ubiquitin protein ligase binding
          identical protein binding
          protein homodimerization activity
          ubiquitin binding
          bHLH transcription factor binding
          sequence-specific DNA binding
          SMAD binding
          metal ion binding
          protein heterodimerization activity
          co-SMAD binding
          R-SMAD binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0900020771.mm.1 TC0900002480.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900020771.mm.1 NM_016769.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900020771.mm.1 NM_016769.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0