Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00490810_m1
          See other SUMO3 GE Assays ›
          Gene Symbol:
          Sumo3
          Gene Name:
          SMT3 suppressor of mif two 3 homolog 3 (yeast)
          Gene Aliases:
          2810014B19Rik, D10Ertd345e, SMT3A, SUMO-3, Smt3h1
          Chromosome Location:
          Chr.10: 77606097 - 77618331 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00490810_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sumo3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001301672.1 NP_001288601.1 1 - 2 241 64
          NM_019929.4 NP_064313.1 1 - 2 241 64
          NR_125904.1 1 - 2 241 64
          GenBank mRNA AF063847.1 - 1 - 2 68 64
          AK012742.1 - 1 - 2 116 64
          AK013019.1 - 1 - 2 110 64
          AK148306.1 - 1 - 2 105 64
          AK150063.1 - 1 - 2 114 64
          AK150200.1 - 1 - 2 114 64
          AK160169.1 - 1 - 2 113 64
          AK162035.1 - 1 - 2 114 64
          AK167207.1 - 1 - 2 82 64
          AK168988.1 - 1 - 2 105 64
          BC115488.1 - 1 - 2 48 40100722 64
          BC115489.1 - 1 - 2 48 40100723 64
          CJ069985.1 - 1 - 2 143 64

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AF063847.1, AK012742.1, AK013019.1, AK148306.1, AK150063.1, AK150200.1, AK160169.1, AK162035.1, AK167207.1, AK168988.1, BC115488.1, BC115489.1, NM_001301672.1, NM_019929.4 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sumo3

          Gene Ontology Categories:

          Function(s) Process(es)

          protein sumoylation
          protein localization to nucleus
          positive regulation of ubiquitin-dependent protein catabolic process
          SUMO transferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000005548.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005548.mm.1 NM_019929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005553.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005553.mm.1 NM_019929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005540.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005540.mm.1 NM_019929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005544.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005544.mm.1 NM_019929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005539.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005539.mm.1 NM_019929.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1000005545.mm.1 TC1000003215.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000005545.mm.1 NM_019929.4 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0