Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491358_g1
          See other F12 GE Assays ›
          Gene Symbol:
          F12
          Gene Name:
          coagulation factor XII (Hageman factor)
          Gene Aliases:
          FXII, HAF
          Chromosome Location:
          Chr.13: 55417966 - 55426802 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491358_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: F12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AW111733.1 - 1 - 2 82
          BC037085.1 - 13 - 14 1639 82
          BC049867.1 - 13 - 14 1662 82
          BC057921.1 - 13 - 14 1650 5101105 82
          X99571.1 - 13 - 14 1645 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300002058.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054919 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054918 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054917 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184949 X99571.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P184949 X99571.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803878 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 374656 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781943 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 462096 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926664 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133327 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P149571 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184949 X99571.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P184949 X99571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184949 X99571.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P184949 X99571.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054971 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054977 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054914 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054917 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054947 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054908 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054913 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054911 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054945 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29054958 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02696937 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P149571 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07882818 - -
          Whole Mouse Genome G4122A A_51_P184949 X99571.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P184949 X99571.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106265 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112396 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P149571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10584504 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10584498 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10584502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054918 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054917 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P29054919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054908 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054904 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29054914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054921 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10662579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054953 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054941 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11063260 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10534091 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10773749 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054951 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10664994 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10584498 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10662593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10457649 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054968 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054950 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054959 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054931 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10569630 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054984 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10593546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054965 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10584499 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054955 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054917 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054914 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10584494 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054946 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10662587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054908 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11036563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10664993 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29054923 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10409452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409455 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409457 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409460 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409462 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409461 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409459 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409463 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409454 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10409458 - -
          Affymetrix HT MG-430 PM Array Plate 1420496_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10409449 - -
          Mouse Genome 430 2.0 Array 1420496_PM_at - -
          Mouse Genome 430 2.0 Array 1420496_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58992_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 58992_at - -
          Affymetrix Mouse Clariom S Array PSR1300012962.mm.1 TC1300002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012962.mm.1 NM_021489.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300012961.mm.1 TC1300002058.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300012961.mm.1 NM_021489.3 Refseq
          Mouse Expression Set 430 1420496_PM_at - -
          Mouse Expression Set 430 1420496_at - -
          Murine Genome U74v2 Set 92509_PM_at - -
          Murine Genome U74v2 Set 92509_at - -
          Mouse Genome 430A 2.0 Array 1420496_PM_at - -
          Mouse Genome 430A 2.0 Array 1420496_at - -
          Affymetrix HT Mouse Genome 430A Array 1420496_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1420496_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1090215 - -
          Illumina mouse-6 v1.0 expression beadchip 1090215 - -
          Illumina mouse-6 v1.0 expression beadchip 3520736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001090215 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2605656 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2746692 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2605656 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2746692 - -
          mouse-6 v1.1 (Illumina) scl43924.13.1_5-S - -
          mouse-6 v1.1 (Illumina) scl0003720.1_47-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520736 - -
          Illumina Sentrix mouse-6.v1 bead array 1090215 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0003720.1_47-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl43924.13.1_5-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1090215 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2605656 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2938668 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2746692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2746692 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2938668 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2605656 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5777fb6d44-4pg6x:80/100.66.134.75:80.
          git-commit: 3a4c0ffdaddae765c0fdfab5c280296089abf14d
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0