Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00491756_m1
          See other SRA1 GE Assays ›
          Gene Symbol:
          Sra1
          Gene Name:
          steroid receptor RNA activator 1
          Gene Aliases:
          AA959952, Sra, Srap, Straa1, Strra1
          Chromosome Location:
          Chr.18: 36667187 - 36670311 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00491756_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sra1
          Interrogated Sequence Translated Protein RNADB ID Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_025291.3 NP_079567.2
          3 - 4 381 75
          NR_028355.1
          2 - 3 499 75
          GenBank mRNA AF092039.1 - LIT1243
          SRA
          2 - 3 285 75
          AK007302.1 -
          3 - 4 367 75
          AK010375.1 -
          3 - 4 367 75
          AK154232.1 -
          3 - 4 351 75
          BC026480.1 -
          1 - 2 160 75
          BC048362.1 -
          3 - 4 353 2646489 75
          BX516162.1 -
          2 - 3 448 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001747.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the BC026480.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sra1

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription by RNA polymerase II
          cell population proliferation
          cell differentiation
          regulation of apoptotic process
          positive regulation of transcription by RNA polymerase II
          regulation of nucleic acid-templated transcription
          positive regulation of signaling receptor activity
          DNA binding
          transcription coregulator activity
          transcription coactivator activity
          nuclear receptor binding
          nuclear receptor coactivator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625184 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 BQ748094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 BC048362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 AK007302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 AK010375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 BQ748094.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 BC048362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 AK007302.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 AK010375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26891 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177915 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P178950 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 BQ748094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 BC048362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 AK007302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 AK010375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 NR_028355.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 BQ748094.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 BC048362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 AK007302.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 AK010375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 BQ748094.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 BC048362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 AK007302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 AK010375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 BQ748094.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 BC048362.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 AK007302.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 AK010375.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625171 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31625179 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P178950 - -
          Whole Mouse Genome G4122A A_51_P226053 BQ748094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 BC048362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 AK007302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 AK010375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 NR_028355.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 BQ748094.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 BC048362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 AK007302.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 AK010375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P226053 NR_028355.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119486 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P178950 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13849106 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14063678 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13849107 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13849098 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31625184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625116 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625131 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31625127 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625176 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625163 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13949364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625172 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14063678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13849111 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13949360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31625169 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10458354 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458353 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458350 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458352 - -
          Affymetrix Mouse Gene 1.0 ST Array 10458351 - -
          Affymetrix HT MG-430 PM Array Plate 1437423_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10458349 - -
          Mouse Genome 430 2.0 Array 1437423_PM_a_at - -
          Mouse Genome 430 2.0 Array 1437423_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24068_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 24068_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800008788.mm.1 TC1800001747.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800008788.mm.1 NM_025291.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008788.mm.1 NM_025291.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008788.mm.1 NR_028355.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800008788.mm.1 NR_028355.1 Refseq
          Mouse Expression Set 430 1437423_a_at - -
          Mouse Expression Set 430 1437423_PM_a_at - -
          Murine Genome U74v2 Set 160323_PM_at - -
          Murine Genome U74v2 Set 160323_at - -
          Mouse Genome 430A 2.0 Array 1437423_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1437423_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437423_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1437423_a_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7000138 - -
          MouseWG-6 v2 Expression BeadChip 3710471 - -
          Illumina mouse-6 v1.0 expression beadchip 3710471 - -
          Illumina mouse-6 v1.0 expression beadchip 7000138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007000138 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003710471 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2659132 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1235049 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2659132 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1235049 - -
          mouse-6 v1.1 (Illumina) scl51510.4.1_91-S - -
          mouse-6 v1.1 (Illumina) scl0002171.1_4-S - -
          Illumina Sentrix mouse-6.v1 bead array 3710471 - -
          Illumina Sentrix mouse-6.v1 bead array 7000138 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51510.4.1_91-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002171.1_4-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7000138 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3710471 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2659132 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1235049 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2876554 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2876554 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2659132 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1235049 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0002171.1_4-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0