Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700005511.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005511.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005511.mm.1 |
NM_001199733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005517.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005517.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005517.mm.1 |
NM_001199733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005517.mm.1 |
NM_007829.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005517.mm.1 |
NM_007829.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005518.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005518.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005518.mm.1 |
NM_001199733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005518.mm.1 |
NM_007829.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005518.mm.1 |
NM_007829.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005519.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005519.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005519.mm.1 |
NM_001199733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005519.mm.1 |
NM_007829.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005519.mm.1 |
NM_007829.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005512.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005512.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005512.mm.1 |
NM_001199733.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005513.mm.1 |
TC1700000600.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700005513.mm.1 |
NM_001199733.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700005513.mm.1 |
NM_001199733.1 |
Refseq |