Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00492996_g1
          See other KRT4 GE Assays ›
          Gene Symbol:
          Krt4
          Gene Name:
          keratin 4
          Gene Aliases:
          AW108092, K4, Krt-2.4, Krt2-4
          Chromosome Location:
          Chr.15: 101918535 - 101924735 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00492996_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Krt4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008475.2 NP_032501.2 5 - 6 1094 80
          GenBank mRNA BC064008.1 - 5 - 6 1075 80
          BC145839.1 - 5 - 6 1049 40131142 80
          BC145841.1 - 5 - 6 1049 40131144 80
          X03491.1 - 5 - 6 1146 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500002278.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Krt4

          Gene Ontology Categories:

          Function(s) Process(es)

          cytoskeleton organization
          embryo development
          epithelial cell differentiation
          negative regulation of epithelial cell proliferation
          structural molecule activity
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P319180 X03491.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P319180 X03491.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 866388 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 508532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 460546 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152160 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P169971 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P319180 X03491.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P319180 X03491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P319180 X03491.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P319180 X03491.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494903 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494886 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494905 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494883 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494861 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494859 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494879 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494894 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494865 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494892 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494868 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30494895 - -
          Whole Mouse Genome G4122A A_51_P319180 X03491.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P319180 X03491.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120447 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P110044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12433133 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12419569 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12433132 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12419570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494861 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30494834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946243 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494828 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494815 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12037653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494819 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12419569 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12037641 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11852179 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12249060 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12419571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11852181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494905 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12013954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12419570 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11946216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494808 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494872 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12037639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494820 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494810 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12433145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12109548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494765 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494851 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494781 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494804 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11882542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12249058 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494875 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494818 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12249061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11882535 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494830 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494802 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494777 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494886 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494793 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11882540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494873 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30494823 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10432889 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432891 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432893 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432894 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432887 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432892 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432888 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432890 - -
          Affymetrix Mouse Gene 1.0 ST Array 10432895 - -
          Affymetrix HT MG-430 PM Array Plate 1418735_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438394_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10432886 - -
          Mouse Genome 430 2.0 Array 1418735_PM_at - -
          Mouse Genome 430 2.0 Array 1418735_at - -
          Mouse Genome 430 2.0 Array 1438394_PM_x_at - -
          Mouse Genome 430 2.0 Array 1438394_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16682_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 16682_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1500018789.mm.1 TC1500002278.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500018789.mm.1 NM_008475.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500018789.mm.1 NM_008475.2 Refseq
          Mouse Expression Set 430 1418735_at - -
          Mouse Expression Set 430 1418735_PM_at - -
          Mouse Expression Set 430 1438394_PM_x_at - -
          Mouse Expression Set 430 1438394_x_at - -
          Murine Genome U74v2 Set 100421_at - -
          Murine Genome U74v2 Set 161647_f_at - -
          Murine Genome U74v2 Set 161647_PM_f_at - -
          Murine Genome U74v2 Set 100421_PM_at - -
          Murine Genome U74v2 Set 167079_PM_f_at - -
          Murine Genome U74v2 Set 167079_f_at - -
          Mouse Genome 430A 2.0 Array 1418735_at - -
          Mouse Genome 430A 2.0 Array 1418735_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418735_at - -
          Affymetrix HT Mouse Genome 430A Array 1418735_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438394_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1438394_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 314757 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2900519 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002900519 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2621930 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2621930 - -
          mouse-6 v1.1 (Illumina) scl46692.9.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 2900519 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl46692.9.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2900519 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2621930 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2621930 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl46692.9.1_22-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0