Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00493469_m1
          See other TFAP2C GE Assays ›
          Gene Symbol:
          Tfap2c
          Gene Name:
          transcription factor AP-2, gamma
          Gene Aliases:
          AA409384, AP2gamma, Ap-2.2, Stra2, Tcfap2c
          Chromosome Location:
          Chr.2: 172545678 - 172558622 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00493469_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tfap2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009335.2 NP_033361.2 1 - 2 291 92
          GenBank mRNA AK145166.1 - 1 - 2 241 92
          BC003778.1 - 1 - 2 165 3592762 92
          BY016798.1 - 1 - 2 292 92
          KJ463836.1 - 1 - 2 87 92
          X94694.1 - 1 - 2 270 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002672.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK145166.1, BC003778.1, KJ463836.1, NM_009335.2, X94694.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tfap2c

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription by RNA polymerase II
          trophectodermal cell differentiation
          hair follicle development
          keratinocyte development
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription by RNA polymerase II
          transcription by RNA polymerase II
          stem cell population maintenance
          forebrain neuron fate commitment
          cerebral cortex development
          cell differentiation
          germ-line stem cell population maintenance
          somatic stem cell population maintenance
          regulation of gene expression, epigenetic
          regulation of cell population proliferation
          skin development
          regulation of epidermis development
          positive regulation of transcription by RNA polymerase II
          sebaceous gland development
          dichotomous subdivision of terminal units involved in mammary gland duct morphogenesis
          epithelial cell proliferation involved in mammary gland duct elongation
          RNA polymerase II transcription regulatory region sequence-specific DNA binding
          RNA polymerase II cis-regulatory region sequence-specific DNA binding
          DNA-binding transcription factor activity, RNA polymerase II-specific
          core promoter sequence-specific DNA binding
          DNA binding
          DNA-binding transcription factor activity
          protein binding
          protein dimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200021742.mm.1 TC0200002672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021742.mm.1 NM_001159696.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021742.mm.1 NM_001159696.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021742.mm.1 NM_009335.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021742.mm.1 NM_009335.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021735.mm.1 TC0200002672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021735.mm.1 NM_009335.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021735.mm.1 NM_009335.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021734.mm.1 TC0200002672.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200021734.mm.1 NM_009335.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200021734.mm.1 NM_009335.2 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 317203 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0