Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00494728_g1
          See other GPAA1 GE Assays ›
          Gene Symbol:
          Gpaa1
          Gene Name:
          GPI anchor attachment protein 1
          Gene Aliases:
          C80044, mGAA1
          Chromosome Location:
          Chr.15: 76331294 - 76334900 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00494728_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gpaa1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010331.2 NP_034461.1 11 - 12 1716 79
          GenBank mRNA AB002136.1 - 11 - 12 1645 79
          AB006970.1 - 11 - 12 1652 79
          AK077518.1 - 11 - 12 1716 79
          AK164219.1 - 11 - 12 1662 79
          AK164978.1 - 11 - 12 1716 79
          AK168057.1 - 11 - 12 1919 79
          BC006697.1 - 11 - 12 1655 3498573 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1500000585.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gpaa1

          Gene Ontology Categories:

          Function(s) Process(es)

          GPI anchor biosynthetic process
          GPI-anchor transamidase activity
          GPI anchor binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347602 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347603 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347595 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347600 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 BC006697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK164219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK164978.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK168057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 BC006697.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK164219.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK164978.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AK168057.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 803650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 166779 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P171190 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 BC006697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK164219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK164978.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK168057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AB006970.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 BC006697.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK164219.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK164978.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AK168057.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 BC006697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK164219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK164978.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK168057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 BC006697.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK164219.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK164978.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AK168057.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347597 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347603 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347604 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347615 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347605 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347602 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347608 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P30347598 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P171190 - -
          Whole Mouse Genome G4122A A_51_P273203 BC006697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK164219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK164978.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK168057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AB006970.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 BC006697.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK164219.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK164978.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AK168057.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P273203 AB006970.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107530 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P122301 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P171190 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12289192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12289186 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12289191 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P12289185 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347602 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347594 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347596 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347601 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347597 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347603 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347600 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347599 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P30347595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347597 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P30347536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347615 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347620 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12078069 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347616 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347606 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12078036 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P30347604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P12078064 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10424812 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424821 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424814 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424816 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424811 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424815 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10424813 - -
          Affymetrix HT MG-430 PM Array Plate 1448225_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1455954_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10424810 - -
          Mouse Genome 430 2.0 Array 1441004_at - -
          Mouse Genome 430 2.0 Array 1448225_PM_at - -
          Mouse Genome 430 2.0 Array 1455954_PM_x_at - -
          Mouse Genome 430 2.0 Array 1441004_PM_at - -
          Mouse Genome 430 2.0 Array 1438152_PM_at - -
          Mouse Genome 430 2.0 Array 1438152_at - -
          Mouse Genome 430 2.0 Array 1455954_x_at - -
          Mouse Genome 430 2.0 Array 1448225_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14731_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14731_at - -
          Affymetrix Mouse Clariom S Array PSR1500004071.mm.1 TC1500000585.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1500004071.mm.1 NM_010331.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1500004071.mm.1 NM_010331.2 Refseq
          Mouse Expression Set 430 1448225_PM_at - -
          Mouse Expression Set 430 1448225_at - -
          Mouse Expression Set 430 1455954_PM_x_at - -
          Mouse Expression Set 430 1455954_x_at - -
          Murine Genome U74v2 Set 161682_f_at - -
          Murine Genome U74v2 Set 161682_PM_f_at - -
          Murine Genome U74v2 Set 100911_at - -
          Murine Genome U74v2 Set 100911_PM_at - -
          Mouse Genome 430A 2.0 Array 1448225_at - -
          Mouse Genome 430A 2.0 Array 1455954_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1455954_x_at - -
          Mouse Genome 430A 2.0 Array 1448225_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1455954_PM_x_at - -
          Affymetrix HT Mouse Genome 430A Array 1448225_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448225_at - -
          Affymetrix HT Mouse Genome 430A Array 1455954_x_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500463 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500463 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255423 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255423 - -
          mouse-6 v1.1 (Illumina) scl47800.9.47_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500463 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl47800.9.47_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500463 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255423 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255423 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0