Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00495374_m1
          See other LNX1 GE Assays ›
          Gene Symbol:
          Lnx1
          Gene Name:
          ligand of numb-protein X 1
          Gene Aliases:
          Lnx
          Chromosome Location:
          Chr.5: 74592447 - 74703042 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00495374_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lnx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001159579.1 NP_001153051.1 1 - 2 369 65
          NM_001159580.1 NP_001153052.1 1 - 2 369 65
          NM_010727.4 NP_034857.3 1 - 2 369 65
          GenBank mRNA AF034746.1 - 1 - 2 297 65
          AK044127.1 - 1 - 2 322 65
          AK138326.1 - 1 - 2 370 65
          BC040367.1 - 1 - 2 321 65
          BC139771.1 - 1 - 2 297 40111506 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0500002608.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lnx1

          Gene Ontology Categories:

          Function(s) Process(es)

          ubiquitin-dependent protein catabolic process
          protein homooligomerization
          ubiquitin-protein transferase activity
          protein binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          PDZ domain binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 TC0500002608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159577.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159577.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159580.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_001159580.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_010727.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023130.mm.1 NM_010727.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 TC0500002608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159580.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_001159580.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_010727.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023133.mm.1 NM_010727.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 TC0500002608.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159578.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159578.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159579.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159579.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159580.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_001159580.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_010727.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0500023135.mm.1 NM_010727.4 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 690253 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0