Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00496713_g1
          See other TCP1 GE Assays ›
          Gene Symbol:
          Tcp1
          Gene Name:
          t-complex protein 1
          Gene Aliases:
          AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63
          Chromosome Location:
          Chr.17: 12916329 - 12925067 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00496713_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tcp1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290712.1 NP_001277641.1 10 - 11 1602 64
          NM_013686.4 NP_038714.2 11 - 12 1688 64
          GenBank mRNA AK149611.1 - 11 - 12 1553 64
          AK149755.1 - 11 - 12 1554 64
          AK150666.1 - 9 - 10 3113 64
          AK150744.1 - 9 - 10 3113 64
          AK151068.1 - 11 - 12 1553 64
          AK151903.1 - 9 - 10 3113 64
          AK152633.1 - 9 - 10 3113 64
          AK152641.1 - 11 - 12 1545 64
          AK152759.1 - 11 - 12 1553 64
          AK153037.1 - 11 - 12 1553 64
          AK153430.1 - 10 - 11 1492 64
          AK165665.1 - 11 - 12 1555 64
          AK166828.1 - 11 - 12 1552 64
          AK166966.1 - 11 - 12 1576 64
          AK167529.1 - 11 - 12 1553 64
          BC003809.1 - 11 - 12 1536 3599670 64
          D90344.1 - 11 - 12 1503 64
          M12899.1 - 11 - 12 1511 64
          M20130.1 - 4 - 5 576 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000143.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tcp1

          Gene Ontology Categories:

          Function(s) Process(es)

          protein folding
          binding of sperm to zona pellucida
          translocation of peptides or proteins into host cell cytoplasm
          toxin transport
          regulation of macrophage apoptotic process
          nucleotide binding
          RNA binding
          protein binding
          ATP binding
          ubiquitin protein ligase binding
          unfolded protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055432 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055431 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 D90344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 M12899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 M20130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 AK166966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 D90344.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 M12899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 M20130.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 AK166966.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619156 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 94414 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 458491 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 523306 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 633965 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 82572 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 D90344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 M12899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 M20130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 AK166966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 AK167529.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 D90344.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 M12899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 M20130.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 AK166966.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 D90344.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 M12899.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 M20130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 AK166966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 D90344.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 M12899.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 M20130.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 AK166966.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055408 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055432 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055404 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055406 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055427 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31055435 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08720822 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08720839 - -
          Whole Mouse Genome G4122A A_52_P4271 AK151903.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P4271 AK152633.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P4271 AK150666.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P4271 AK150744.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 D90344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 M12899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 M20130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 AK166966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 AK167529.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 D90344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 M12899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 M20130.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 AK166966.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P164030 AK167529.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138664 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08720839 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457704 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457701 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457685 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13457703 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055430 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055431 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31055432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055408 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055390 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31055407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055402 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457706 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055379 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055415 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055384 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13062637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055410 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457698 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13457703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055413 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055418 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055429 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055424 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055414 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13413426 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055390 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055380 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055400 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055382 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055404 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13067173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055396 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055395 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055408 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055420 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31055405 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10441801 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441798 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441809 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441808 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441810 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441799 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441805 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441807 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441803 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441800 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441806 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441802 - -
          Affymetrix Mouse Gene 1.0 ST Array 10441804 - -
          Affymetrix HT MG-430 PM Array Plate 1448122_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10447885 - -
          Affymetrix Mouse Gene 1.1 ST Array 10441797 - -
          Mouse Genome 430 2.0 Array 1448122_at - -
          Mouse Genome 430 2.0 Array 1448122_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21454_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 21454_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700001121.mm.1 TC1700000143.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001121.mm.1 NM_013686.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001118.mm.1 TC1700000143.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001118.mm.1 NM_013686.4 Refseq
          Mouse Expression Set 430 1448122_at - -
          Mouse Expression Set 430 1448122_PM_at - -
          Murine Genome U74v2 Set 94218_at - -
          Murine Genome U74v2 Set 162443_PM_at - -
          Murine Genome U74v2 Set 162443_at - -
          Murine Genome U74v2 Set 94218_PM_at - -
          Murine Genome U74v2 Set 133457_PM_at - -
          Murine Genome U74v2 Set 133457_at - -
          Mouse Genome 430A 2.0 Array 1448122_at - -
          Mouse Genome 430A 2.0 Array 1448122_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1448122_at - -
          Affymetrix HT Mouse Genome 430A Array 1448122_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2450100 - -
          Illumina mouse-6 v1.0 expression beadchip 5130524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005130524 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450100 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2677386 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774140 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2735977 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2735977 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2677386 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774140 - -
          mouse-6 v1.1 (Illumina) scl51111.10.24_9-S - -
          mouse-6 v1.1 (Illumina) scl0001621.1_13-S - -
          Illumina Sentrix mouse-6.v1 bead array 2450100 - -
          Illumina Sentrix mouse-6.v1 bead array 5130524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0001621.1_13-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51111.10.24_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5130524 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450100 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2774140 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2677386 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774140 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2677386 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0001621.1_13-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51111.10.24_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0