Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00497383_g1
          See other CAPRIN1 GE Assays ›
          Gene Symbol:
          Caprin1
          Gene Name:
          cell cycle associated protein 1
          Gene Aliases:
          AL022980, Caprin-1, Gpiap, Gpiap1, Mmgpip137, P137, rng105
          Chromosome Location:
          Chr.2: 103762945 - 103797640 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00497383_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Caprin1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001111289.1 NP_001104759.1 7 - 8 1016 68
          NM_001111290.1 NP_001104760.1 7 - 8 1016 68
          NM_001111291.1 NP_001104761.1 8 - 9 1056 68
          NM_001111292.1 NP_001104762.1 7 - 8 1016 68
          NM_016739.3 NP_058019.2 8 - 9 1056 68
          GenBank mRNA AB373955.1 - 7 - 8 878 68
          AK141992.1 - 7 - 8 958 68
          AK147514.1 - 7 - 8 2827 68
          AK148203.1 - 7 - 8 1003 68
          AK157707.1 - 8 - 9 1040 68
          AK161131.1 - 2 - 3 151 68
          AK166461.1 - 7 - 8 974 68
          AK168676.1 - 7 - 8 1011 68
          AK170830.1 - 8 - 9 1018 68
          AK193449.1 - 1 - 2 46 68
          BC052427.1 - 8 - 9 1035 6836041 68
          U27838.1 - 7 - 8 993 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200004229.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Caprin1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of translation
          cell differentiation
          positive regulation of dendrite morphogenesis
          positive regulation of dendritic spine morphogenesis
          RNA binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 TC0200004229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111289.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111289.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111290.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111290.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111291.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111291.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_001111292.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_016739.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035593.mm.1 NM_016739.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 TC0200004229.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111289.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111289.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111290.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111290.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111291.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111291.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111292.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_001111292.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_016739.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200035592.mm.1 NM_016739.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0