Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498085_m1
          See other CYB561D2 GE Assays ›
          Gene Symbol:
          Cyb561d2
          Gene Name:
          cytochrome b-561 domain containing 2
          Gene Aliases:
          AW049681, Tsp10
          Chromosome Location:
          Chr.9: 107539005 - 107542188 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498085_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyb561d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_019720.4 NP_062694.1 2 - 3 410 58
          GenBank mRNA AF131206.1 - 3 - 4 282 58
          AK030225.1 - 2 - 3 411 58
          AK162586.1 - 2 - 3 397 58
          BC027212.1 - 3 - 4 279 3993231 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900003033.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyb561d2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          molecular_function
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 BC027212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 NM_019720.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AF131206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AK030225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 BC027212.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 NM_019720.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AF131206.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AK030225.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 488344 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154073 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P167625 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P132695 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 BC027212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 NM_019720.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AF131206.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AK030225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AK162586.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 BC027212.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 NM_019720.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AF131206.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AK030225.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 BC027212.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 NM_019720.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AF131206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AK030225.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 BC027212.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 NM_019720.4 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AF131206.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AK030225.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809409 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809394 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26809403 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P167625 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P132695 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154073 - -
          Whole Mouse Genome G4122A A_51_P103509 BC027212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 NM_019720.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AF131206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AK030225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AK162586.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 BC027212.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 NM_019720.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AF131206.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AK030225.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P103509 AK162586.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123375 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P132695 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P167625 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154073 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07487490 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07487469 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07487465 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07487467 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07487487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809350 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26809413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07487481 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07487471 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809401 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809416 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07758575 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809405 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26809398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07487476 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10596665 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596670 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596668 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596669 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596666 - -
          Affymetrix Mouse Gene 1.0 ST Array 10596667 - -
          Affymetrix HT MG-430 PM Array Plate 1418482_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10596664 - -
          Mouse Genome 430 2.0 Array 1418482_at - -
          Mouse Genome 430 2.0 Array 1418482_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56368_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 56368_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0900025322.mm.1 TC0900003033.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025322.mm.1 NM_019720.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025322.mm.1 NM_019720.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025323.mm.1 TC0900003033.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900025323.mm.1 NM_019720.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900025323.mm.1 NM_019720.4 Refseq
          Mouse Expression Set 430 1418482_at - -
          Mouse Expression Set 430 1418482_PM_at - -
          Mouse Genome 430A 2.0 Array 1418482_PM_at - -
          Mouse Genome 430A 2.0 Array 1418482_at - -
          Affymetrix HT Mouse Genome 430A Array 1418482_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418482_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 568904 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 460671 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000460671 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2775390 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2775390 - -
          mouse-6 v1.1 (Illumina) scl35375.4_95-S - -
          Illumina Sentrix mouse-6.v1 bead array 460671 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl35375.4_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460671 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2850342 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2850342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0