Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498536_m1
          See other PARK7 GE Assays ›
          Gene Symbol:
          Park7
          Gene Name:
          Parkinson disease (autosomal recessive, early onset) 7
          Gene Aliases:
          DJ-1, Dj1
          Chromosome Location:
          Chr.4: 150897133 - 150914432 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498536_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Park7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020569.3 NP_065594.2 1 - 2 144 93
          GenBank mRNA AB015652.1 - 1 - 2 91 93
          AK153948.1 - 1 - 2 101 93
          AK168341.1 - 1 - 2 74 93
          CA456587.1 - 1 - 2 182 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400004070.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AB015652.1, AK153948.1, AK168341.1, NM_020569.3 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Park7

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of protein phosphorylation
          synaptic transmission, dopaminergic
          negative regulation of protein kinase activity
          proteolysis
          protein deglycosylation
          autophagy
          inflammatory response
          response to oxidative stress
          mitochondrion organization
          single fertilization
          cell population proliferation
          adult locomotory behavior
          detoxification of copper ion
          positive regulation of gene expression
          negative regulation of gene expression
          methylglyoxal catabolic process to D-lactate via S-lactoyl-glutathione
          lactate biosynthetic process
          negative regulation of protein ubiquitination
          negative regulation of protein binding
          negative regulation of proteasomal ubiquitin-dependent protein catabolic process
          positive regulation of interleukin-8 production
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of protein sumoylation
          cellular response to oxidative stress
          cellular response to reactive oxygen species
          cellular response to glyoxal
          negative regulation of protein catabolic process
          response to hydrogen peroxide
          hydrogen peroxide metabolic process
          negative regulation of apoptotic process
          regulation of neuron apoptotic process
          negative regulation of neuron apoptotic process
          positive regulation of transcription by RNA polymerase II
          glycolate biosynthetic process
          negative regulation of protein export from nucleus
          regulation of inflammatory response
          detoxification of mercury ion
          protein stabilization
          positive regulation of DNA-binding transcription factor activity
          negative regulation of ubiquitin-protein transferase activity
          dopamine uptake involved in synaptic transmission
          regulation of mitochondrial membrane potential
          membrane depolarization
          membrane hyperpolarization
          negative regulation of cell death
          regulation of androgen receptor signaling pathway
          cellular response to hydrogen peroxide
          positive regulation of protein homodimerization activity
          positive regulation of protein localization to nucleus
          negative regulation of neuron death
          positive regulation of superoxide dismutase activity
          negative regulation of protein acetylation
          positive regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          positive regulation of mitochondrial electron transport, NADH to ubiquinone
          negative regulation of protein K48-linked deubiquitination
          negative regulation of TRAIL-activated apoptotic signaling pathway
          positive regulation of pyrroline-5-carboxylate reductase activity
          positive regulation of tyrosine 3-monooxygenase activity
          positive regulation of dopamine biosynthetic process
          glyoxal catabolic process
          positive regulation of L-dopa biosynthetic process
          positive regulation of L-dopa decarboxylase activity
          negative regulation of oxidative stress-induced cell death
          negative regulation of oxidative stress-induced neuron death
          negative regulation of hydrogen peroxide-induced cell death
          negative regulation of hydrogen peroxide-induced neuron death
          negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of hydrogen peroxide-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of reactive oxygen species biosynthetic process
          negative regulation of ubiquitin-specific protease activity
          positive regulation of oxidative phosphorylation uncoupler activity
          positive regulation of transcription regulatory region DNA binding
          positive regulation of androgen receptor activity
          negative regulation of extrinsic apoptotic signaling pathway
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          transcription coactivator activity
          RNA binding
          mRNA binding
          histone deacetylase activity
          signaling receptor binding
          copper ion binding
          protein binding
          transcription factor binding
          peptidase activity
          protein-N-terminal asparagine amidohydrolase activity
          UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
          CoA hydrolase activity
          superoxide dismutase copper chaperone activity
          oxidoreductase activity, acting on peroxide as acceptor
          hydrolase activity
          iprodione amidohydrolase activity
          (3,5-dichlorophenylurea)acetate amidohydrolase activity
          glyoxalase III activity
          enzyme binding
          kinase binding
          cytokine binding
          4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
          didemethylisoproturon amidohydrolase activity
          N-isopropylacetanilide amidohydrolase activity
          N-cyclohexylformamide amidohydrolase activity
          2-oxoglutaryl-CoA thioesterase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA thioesterase activity
          isonicotinic acid hydrazide hydrolase activity
          cis-aconitamide amidase activity
          gamma-N-formylaminovinylacetate hydrolase activity
          3-isopropylbut-3-enoyl-CoA thioesterase activity
          tyrosine 3-monooxygenase activator activity
          L-dopa decarboxylase activator activity
          identical protein binding
          protein homodimerization activity
          O-succinylbenzoate synthase activity
          indoleacetamide hydrolase activity
          N-acetylcitrulline deacetylase activity
          small protein activating enzyme binding
          ubiquitin-like protein conjugating enzyme binding
          glutaryl-CoA hydrolase activity
          mercury ion binding
          N-acetylgalactosamine-6-phosphate deacetylase activity
          acyl-CoA hydrolase activity
          androgen receptor binding
          peroxiredoxin activity
          diacetylchitobiose deacetylase activity
          chitooligosaccharide deacetylase activity
          scaffold protein binding
          DNA-binding transcription factor binding
          cupric ion binding
          cuprous ion binding
          ubiquitin-specific protease binding
          glyoxalase (glycolic acid-forming) activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400033834.mm.1 TC0400004070.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033834.mm.1 NM_020569.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033834.mm.1 NM_020569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033844.mm.1 TC0400004070.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033844.mm.1 NM_020569.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033844.mm.1 NM_020569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033842.mm.1 TC0400004070.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033842.mm.1 NM_020569.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033842.mm.1 NM_020569.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033845.mm.1 TC0400004070.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033845.mm.1 NM_020569.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033845.mm.1 NM_020569.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 537632 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0