Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00498553_m1
          See other NELFCD GE Assays ›
          Gene Symbol:
          Nelfcd
          Gene Name:
          negative elongation factor complex member C/D, Th1l
          Gene Aliases:
          2410003I03Rik, C77797, Th1, Th1l
          Chromosome Location:
          Chr.2: 174415804 - 174427503 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00498553_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nelfcd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020580.3 NP_065605.2 1 - 2 148 77
          GenBank mRNA AJ238741.1 - 1 - 2 82 77
          AJ238742.1 - 1 - 2 148 77
          AJ238743.1 - 1 - 2 102 77
          AK010360.1 - 1 - 2 95 77
          AK049418.1 - 1 - 2 120 77
          AK159866.1 - 1 - 2 101 77
          BC031747.1 - 1 - 2 106 3673038 77

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200002705.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nelfcd

          Gene Ontology Categories:

          Function(s)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          negative regulation of transcription, DNA-templated

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875216 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875215 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982156 BC031747.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982156 AK049418.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982156 AK159866.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982156 AJ238742.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1982156 AJ238743.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 323892 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 693675 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 525485 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 169484 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 145258 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 343312 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 284387 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 115821 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P131617 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P148043 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982156 BC031747.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982156 AK049418.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982156 AK159866.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982156 AJ238742.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1982156 AJ238743.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875201 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875222 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875229 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21875225 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P148043 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P131617 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04796988 - -
          Whole Mouse Genome G4122A A_51_P126817 BC006959.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126817 BC031747.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126817 AK049418.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126817 AK159866.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P126817 AK182217.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P138717 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P131617 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P148043 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02212826 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01326194 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02212828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01326183 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875212 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875214 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875215 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21875216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875371 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21875220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376794 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875265 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875201 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428146 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428116 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875251 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428132 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875248 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01717226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875220 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01538512 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02212825 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875226 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875249 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01326183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02212826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01329426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01428111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01846664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875204 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875263 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01326194 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02069005 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02259267 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875253 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21875218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01376792 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10479149 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479140 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479137 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479141 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479138 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479152 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479147 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479150 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479142 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479148 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479145 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479153 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479146 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479144 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479151 - -
          Affymetrix Mouse Gene 1.0 ST Array 10479139 - -
          Affymetrix HT MG-430 PM Array Plate 1416198_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10479136 - -
          Mouse Genome 430 2.0 Array 1416198_PM_at - -
          Mouse Genome 430 2.0 Array 1416198_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57314_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 57314_at - -
          Affymetrix Mouse Clariom S Array PSR0200022125.mm.1 TC0200002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022125.mm.1 NM_020580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022125.mm.1 NM_020580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022126.mm.1 TC0200002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022126.mm.1 NM_020580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022126.mm.1 NM_020580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022122.mm.1 TC0200002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022122.mm.1 NM_020580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022122.mm.1 NM_020580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022129.mm.1 TC0200002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022129.mm.1 NM_020580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022129.mm.1 NM_020580.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022128.mm.1 TC0200002705.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200022128.mm.1 NM_020580.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200022128.mm.1 NM_020580.3 Refseq
          Mouse Expression Set 430 1416198_at - -
          Mouse Expression Set 430 1416198_PM_at - -
          Murine Genome U74v2 Set 94015_PM_at - -
          Murine Genome U74v2 Set 94015_at - -
          Mouse Genome 430A 2.0 Array 1416198_PM_at - -
          Mouse Genome 430A 2.0 Array 1416198_at - -
          Affymetrix HT Mouse Genome 430A Array 1416198_at - -
          Affymetrix HT Mouse Genome 430A Array 1416198_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 826385 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1190632 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001190632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2606873 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2606873 - -
          mouse-6 v1.1 (Illumina) scl19818.14.1_56-S - -
          Illumina Sentrix mouse-6.v1 bead array 1190632 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19818.14.1_56-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1190632 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2606873 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2606873 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0