Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499502_m1
          See other LIMS1 GE Assays ›
          Gene Symbol:
          Lims1
          Gene Name:
          LIM and senescent cell antigen-like domains 1
          Gene Aliases:
          2310016J22Rik, 4921524A02Rik, AI507642, AU021743, AW551584, C430041B13Rik, Lims1l, PINCH, PINCH1
          Chromosome Location:
          Chr.10: 58323435 - 58424691 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499502_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lims1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001193303.1 NP_001180232.1 3 - 4 455 93
          NM_026148.3 NP_080424.2 3 - 4 449 93
          NM_201242.1 NP_957694.1 3 - 4 473 93
          GenBank mRNA AK009390.1 - 3 - 4 456 93
          AK014954.1 - 3 - 4 450 93
          AK082932.1 - 3 - 4 318 93
          AK088939.1 - 3 - 4 324 93
          AK151938.1 - 3 - 4 316 93
          AK152679.1 - 3 - 4 316 93
          AK153346.1 - 3 - 4 324 93
          BC005621.1 - 3 - 4 304 3711611 93
          BC067393.1 - 3 - 4 247 93
          DQ003608.1 - 3 - 4 440 93
          EU122953.1 - 3 - 4 439 93
          EU122954.1 - 3 - 4 307 93

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000000514.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lims1

          Gene Ontology Categories:

          Function(s) Process(es)

          cell-matrix adhesion
          establishment or maintenance of cell polarity
          positive regulation of gene expression
          positive regulation of cell-substrate adhesion
          chordate embryonic development
          negative regulation of apoptotic process
          positive regulation of GTPase activity
          establishment of protein localization
          cell-cell junction organization
          negative regulation of transcription, DNA-templated
          negative regulation of epithelial cell proliferation
          protein heterooligomerization
          positive regulation of focal adhesion assembly
          cell-cell adhesion
          positive regulation of substrate adhesion-dependent cell spreading
          negative regulation of neural precursor cell proliferation
          negative regulation of hepatocyte proliferation
          positive regulation of integrin-mediated signaling pathway
          protein binding
          zinc ion binding
          protein kinase binding
          metal ion binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 TC1000000514.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_001193303.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_001193303.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_026148.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_026148.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_201242.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000003408.mm.1 NM_201242.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0