Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00499663_g1
          See other CXXC1 GE Assays ›
          Gene Symbol:
          Cxxc1
          Gene Name:
          CXXC finger 1 (PHD domain)
          Gene Aliases:
          2410002I16Rik, 5830420C16Rik, AI426635, Cfp1, Cgbp, PHF18
          Chromosome Location:
          Chr.18: 74216093 - 74221496 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00499663_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cxxc1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028868.3 NP_083144.1 14 - 15 2102 76
          GenBank mRNA AK010337.1 - 14 - 15 2103 76
          AK083655.1 - 14 - 15 2107 76
          AK146688.1 - 14 - 15 2186 76
          AY099096.1 - 14 - 15 2074 76
          BC012261.1 - 10 - 11 1645 76
          BC030938.1 - 14 - 15 1934 3980339 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000787.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cxxc1

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          histone H3-K4 methylation
          cis-regulatory region sequence-specific DNA binding
          DNA binding
          zinc ion binding
          histone methyltransferase activity (H3-K4 specific)
          unmethylated CpG binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830236 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830235 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 BC030938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AY099096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 BC012261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AK083655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 BC030938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AY099096.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 BC012261.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AK083655.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 853275 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 847857 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899036 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P153300 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 BC030938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AY099096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 BC012261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AK083655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AK146688.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 BC030938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AY099096.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 BC012261.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AK083655.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 BC030938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AY099096.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 BC012261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AK083655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 BC030938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AY099096.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 BC012261.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AK083655.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830232 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830244 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830238 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31830255 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03513593 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P153300 - -
          Whole Mouse Genome G4122A A_51_P262533 BC030938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AY099096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 BC012261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AK083655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AK146688.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 BC030938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AY099096.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 BC012261.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AK083655.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P262533 AK146688.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P101243 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P153300 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14051589 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13760300 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830235 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830232 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31830236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31830289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830239 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13953590 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830264 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13702668 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830242 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830253 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830268 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830290 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13760302 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830221 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830241 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14051596 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830267 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830220 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830271 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13702676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830277 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13702678 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13909435 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830234 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830232 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830265 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830225 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830257 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13721549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830250 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13702670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13721532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830237 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13909432 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31830238 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10456604 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456618 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456615 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456603 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456611 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456613 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456614 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456616 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456609 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456612 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456610 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456617 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456602 - -
          Affymetrix Mouse Gene 1.0 ST Array 10456607 - -
          Affymetrix HT MG-430 PM Array Plate 1452221_PM_a_at - -
          Affymetrix HT MG-430 PM Array Plate 1454106_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10456601 - -
          Mouse Genome 430 2.0 Array 1454106_a_at - -
          Mouse Genome 430 2.0 Array 1454106_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452221_PM_a_at - -
          Mouse Genome 430 2.0 Array 1452221_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74322_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74322_at - -
          Affymetrix Mouse Clariom S Array PSR1800005644.mm.1 TC1800000787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005644.mm.1 NM_028868.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005644.mm.1 NM_028868.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005645.mm.1 TC1800000787.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005645.mm.1 NM_028868.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005645.mm.1 NM_028868.3 Refseq
          Mouse Expression Set 430 1452221_a_at - -
          Mouse Expression Set 430 1452221_PM_a_at - -
          Murine Genome U74v2 Set 103099_PM_f_at - -
          Murine Genome U74v2 Set 103099_f_at - -
          Murine Genome U74v2 Set 169024_f_at - -
          Murine Genome U74v2 Set 170358_f_at - -
          Murine Genome U74v2 Set 170358_PM_f_at - -
          Murine Genome U74v2 Set 169024_PM_f_at - -
          Mouse Genome 430A 2.0 Array 1452221_a_at - -
          Mouse Genome 430A 2.0 Array 1452221_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454106_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452221_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1454106_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1452221_a_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3940136 - -
          Illumina mouse-6 v1.0 expression beadchip 630170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000630170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2706251 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2620959 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2706251 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2620959 - -
          mouse-6 v1.1 (Illumina) scl0002160.1_0-S - -
          mouse-6 v1.1 (Illumina) scl51774.11.3_64-S - -
          Illumina Sentrix mouse-6.v1 bead array 630170 - -
          Illumina Sentrix mouse-6.v1 bead array 3940136 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl51774.11.3_64-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0002160.1_0-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940136 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 630170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2898135 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2620959 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2706251 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2620959 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2898135 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2706251 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl51774.11.3_64-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7699c4d8ff-7jgns:80/100.66.130.62:80.
          git-commit: c11338b6b8d815146c5ef25758b63739976b60cb
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline