Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Baiap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037754.3 | NP_001032843.1 | 2 - 3 | 230 | 72 | |
NM_001037755.3 | NP_001032844.2 | 2 - 3 | 230 | 72 | ||
NM_130862.4 | NP_570932.2 | 2 - 3 | 230 | 72 | ||
GenBank mRNA | AF390178.1 | - | 2 - 3 | 188 | 72 | |
AF390179.1 | - | 2 - 3 | 188 | 72 | ||
AK049469.1 | - | 2 - 3 | 231 | 72 | ||
AK145924.1 | - | 2 - 3 | 231 | 72 | ||
AK158579.1 | - | 2 - 3 | 255 | 72 | ||
AK160401.1 | - | 2 - 3 | 231 | 72 | ||
BC006620.1 | - | 2 - 3 | 221 | 3158004 | 72 | |
BC016411.1 | - | 2 - 3 | 221 | 3158004 | 72 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |