Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502028_m1
          See other ZSCAN12 GE Assays ›
          Gene Symbol:
          Zscan12
          Gene Name:
          zinc finger and SCAN domain containing 12
          Gene Aliases:
          2510038J07Rik, FPM315, Zfp96, mKIAA0426
          Chromosome Location:
          Chr.13: 21362820 - 21372302 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502028_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zscan12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_016684.2 NP_057893.2 3 - 4 674 72
          GenBank mRNA AK075886.1 - 3 - 4 668 72
          AK129138.1 - 3 - 4 674 72
          AK139837.1 - 4 - 5 997 72
          AK142005.1 - 3 - 4 653 72
          BC027041.1 - 2 - 3 632 5364568 72
          U62908.1 - 2 - 3 983 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000164.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zscan12

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          biological_process
          nucleic acid binding
          DNA binding
          DNA-binding transcription factor activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872463 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872465 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872464 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872462 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158552 U62908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P158552 U62908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008176 BC027041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008176 U62908.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008176 AK139837.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008176 AK142005.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2008176 NM_016684.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 544955 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936991 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 366853 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 718456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 218524 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 281172 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102601 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166916 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158552 U62908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P158552 U62908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008176 BC027041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008176 U62908.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008176 AK139837.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008176 AK142005.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2008176 NM_016684.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158552 U62908.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P158552 U62908.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872476 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872478 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872477 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872463 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872464 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872484 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872480 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872459 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872483 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872481 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872452 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872461 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872482 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872457 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872458 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872467 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872466 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872455 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872446 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872444 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872470 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872475 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28872472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02643939 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166916 - -
          Whole Mouse Genome G4122A A_51_P158552 U62908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P158552 U62908.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393131 BP759768.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393131 NM_016684.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393131 AK129138.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P393131 AK135506.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P120360 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123261 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P02643939 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166916 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101466 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101460 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101461 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11107927 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101473 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101472 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101477 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P11101476 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872462 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872466 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872464 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872463 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28872465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872480 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872489 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872496 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872437 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872440 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872475 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872491 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872493 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872457 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872483 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872467 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872510 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872505 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872470 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872501 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28872433 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872488 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872466 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10914696 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10499694 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10758727 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872476 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10465019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872439 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872496 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10836352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10666768 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872491 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872510 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872430 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872497 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10689104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872444 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872467 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11101470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872445 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872461 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11074372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11107927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872449 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872437 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10836353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10592579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872450 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10592578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872503 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10619736 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872493 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872512 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872472 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28872479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10776160 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403924 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403923 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403922 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403921 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403920 - -
          Affymetrix HT MG-430 PM Array Plate 1444149_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451281_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10403919 - -
          Mouse Genome 430 2.0 Array 1444149_PM_at - -
          Mouse Genome 430 2.0 Array 1451281_PM_at - -
          Mouse Genome 430 2.0 Array 1451281_at - -
          Mouse Genome 430 2.0 Array 1444149_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22758_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 22758_at - -
          Affymetrix Mouse Clariom S Array PSR1300001126.mm.1 TC1300000164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001126.mm.1 NM_016684.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001126.mm.1 NM_016684.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001128.mm.1 TC1300000164.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300001128.mm.1 NM_016684.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300001128.mm.1 NM_016684.2 Refseq
          Mouse Expression Set 430 1451281_PM_at - -
          Mouse Expression Set 430 1451281_at - -
          Mouse Expression Set 430 1444149_PM_at - -
          Mouse Expression Set 430 1444149_at - -
          Murine Genome U74v2 Set 104604_at - -
          Murine Genome U74v2 Set 104604_PM_at - -
          Mouse Genome 430A 2.0 Array 1451281_at - -
          Mouse Genome 430A 2.0 Array 1451281_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451281_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451281_at - -
          Affymetrix HT Mouse Genome 430B Array 1444149_at - -
          Affymetrix HT Mouse Genome 430B Array 1444149_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 924325 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4670110 - -
          Illumina mouse-6 v1.0 expression beadchip 4670110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004670110 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1246450 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1246450 - -
          mouse-6 v1.1 (Illumina) scl0022758.1_226-S - -
          Illumina Sentrix mouse-6.v1 bead array 4670110 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0022758.1_226-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4670110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1246450 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1246450 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0022758.1_226-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-9d5985c9f-cd4wl:80/100.66.130.62:80.
          git-commit: 2310f86ea667df8f532fad8d69a2d0b8b05e2a1e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0