Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502277_m1
          See other SAE1 GE Assays ›
          Gene Symbol:
          Sae1
          Gene Name:
          SUMO1 activating enzyme subunit 1
          Gene Aliases:
          2400010M20Rik, 2610044L12Rik, AL033372, AOS1, AW743391, D7Ertd177e, HSPC140, Sua1, Uble1a
          Chromosome Location:
          Chr.7: 16320236 - 16387896 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502277_m1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sae1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001285891.1 NP_001272820.1 1 - 2 256 94
          NM_001285892.1 NP_001272821.1 1 - 2 256 94
          NM_019748.3 NP_062722.1 1 - 2 256 94
          GenBank mRNA AB024303.1 - 1 - 2 144 94
          AK010313.1 - 1 - 2 133 94
          AK011772.1 - 1 - 2 167 94
          AK087556.1 - 1 - 2 147 94
          AK090012.1 - 1 - 2 143 94
          AK154139.1 - 1 - 2 169 94
          AK159672.1 - 1 - 2 158 94
          AK162789.1 - 1 - 2 144 94
          BB838763.1 - 1 - 2 257 94
          BC068164.1 - 1 - 2 145 30359189 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002426.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sae1

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular protein modification process
          regulation of mitotic cell cycle
          protein sumoylation
          positive regulation of protein targeting to mitochondrion
          protein C-terminus binding
          ubiquitin-like modifier activating enzyme activity
          ligase activity
          SUMO activating enzyme activity
          ATP-dependent protein binding
          protein heterodimerization activity

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0700021391.mm.1 TC0700002426.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021391.mm.1 NM_001285891.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021391.mm.1 NM_001285892.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021391.mm.1 NM_019748.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021393.mm.1 TC0700002426.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021393.mm.1 NM_001285891.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021393.mm.1 NM_001285892.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021393.mm.1 NM_019748.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021390.mm.1 TC0700002426.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021390.mm.1 NM_001285891.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021390.mm.1 NM_001285892.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021390.mm.1 NM_019748.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 930840 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0