Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00502938_g1
          See other RPS4L GE Assays ›
          Gene Symbol:
          Rps4l
          Gene Name:
          ribosomal protein S4-like
          Gene Aliases:
          1110033J19Rik, AA407494, Rps4y2
          Chromosome Location:
          Chr.6: 148354656 - 148355596 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00502938_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rps4l
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_003634.2 1 864 60
          GenBank mRNA AK004068.1 - 1 865 60
          AK012382.1 - 1 864 60
          AK132160.1 - 1 865 60
          BC047994.1 - 1 836 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001628 NR_003634.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001628 AK004068.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001628 AK132160.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2001628 BC047994.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001628 NR_003634.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001628 AK004068.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001628 AK132160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2001628 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 NR_003634.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK004068.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK012382.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 AK132160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P406105 BC047994.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10542873 - -
          Affymetrix Mouse Gene 1.0 ST Array 10567107 - -
          Affymetrix Mouse Gene 1.0 ST Array 10542874 - -
          Affymetrix HT MG-430 PM Array Plate 1452730_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10567106 - -
          Affymetrix Mouse Gene 1.1 ST Array 10542872 - -
          Mouse Genome 430 2.0 Array 1452730_at - -
          Mouse Genome 430 2.0 Array 1452730_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66184_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66184_at - -
          Mouse Expression Set 430 1452730_PM_at - -
          Mouse Expression Set 430 1452730_at - -
          Murine Genome U74v2 Set 98149_PM_s_at - -
          Murine Genome U74v2 Set 98148_at - -
          Murine Genome U74v2 Set 98148_PM_at - -
          Murine Genome U74v2 Set 98149_s_at - -
          Mouse Genome 430A 2.0 Array 1452730_PM_at - -
          Mouse Genome 430A 2.0 Array 1452730_at - -
          Affymetrix HT Mouse Genome 430A Array 1452730_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1452730_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 892320 - -
          ABI Mouse Genome Survey Microarray 896629 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2737090 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2737090 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-998b77544-nf2nv:80/100.66.134.75:80.
          git-commit: efd2b0e08ab99238c7874a09efb06372d741ac0a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc