Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503219_g1
          See other CUTA GE Assays ›
          Gene Symbol:
          Cuta
          Gene Name:
          cutA divalent cation tolerance homolog (E. coli)
          Gene Aliases:
          0610039D01Rik, 1810022E02Rik, 1810060C03Rik, 2700094G22Rik, AI326454
          Chromosome Location:
          Chr.17: 26937972 - 26939815 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503219_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cuta
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026307.3 NP_080583.3 6 - 7 503 63
          NM_026948.3 NP_081224.1 5 - 6 451 63
          NR_104458.1 5 - 6 537 63
          GenBank mRNA AK002828.1 - 6 - 7 447 63
          AK003381.1 - 5 - 6 391 63
          AK003647.1 - 6 - 7 445 63
          AK007580.1 - 4 - 5 580 63
          AK012608.1 - 3 - 4 923 63
          AK207031.1 - 3 - 4 110 63
          AK207668.1 - 2 - 3 185 63
          BC021463.1 - 4 - 5 565 63
          BC024422.1 - 6 - 7 433 5005082 63
          BC028531.1 - 5 - 6 542 63
          BC037500.1 - 3 - 4 533 63
          CA323205.1 - 5 - 6 417 63
          CK624897.1 - 5 - 6 428 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700001711.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cuta

          Gene Ontology Categories:

          Function(s) Process(es)

          protein localization
          response to metal ion
          protein homooligomerization
          enzyme binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113297 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113352 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113296 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174905 BC028531.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174905 BC037500.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174905 BC021463.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174905 BC024422.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2174905 AK012608.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 901674 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165711 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174905 BC028531.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174905 BC037500.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174905 BC021463.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174905 BC024422.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2174905 AK012608.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113338 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113335 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113315 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113282 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31113295 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165711 - -
          Whole Mouse Genome G4122A A_51_P373441 BC028531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373441 BC037500.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373441 BC021463.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373441 BC024422.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P373441 AK012608.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P140458 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115974 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165711 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064188 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064192 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064183 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064209 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064198 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064199 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064200 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13064207 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113352 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113295 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113294 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113296 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113292 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113293 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113353 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113354 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113298 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113369 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31113297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113354 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113282 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113338 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113244 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113341 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31113330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13567645 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113337 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113320 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113297 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13513284 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113328 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295108 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113295 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113289 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113286 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113335 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13064200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113349 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113305 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113347 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113341 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113373 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113285 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113344 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113283 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13064188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113292 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113316 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113361 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113359 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113322 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13232911 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13201146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13295107 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13064198 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113332 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13567629 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113308 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113287 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113357 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13064191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31113300 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10449299 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449301 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449300 - -
          Affymetrix Mouse Gene 1.0 ST Array 10449302 - -
          Affymetrix HT MG-430 PM Array Plate 1451387_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10449295 - -
          Mouse Genome 430 2.0 Array 1451387_s_at - -
          Mouse Genome 430 2.0 Array 1451387_PM_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67675_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 67675_at - -
          Affymetrix Mouse Clariom S Array PSR1700015825.mm.1 TC1700001711.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700015825.mm.1 NM_026307.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015825.mm.1 NM_026948.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700015825.mm.1 NR_104458.1 Refseq
          Mouse Expression Set 430 1451387_s_at - -
          Mouse Expression Set 430 1451387_PM_s_at - -
          Murine Genome U74v2 Set 95113_at - -
          Murine Genome U74v2 Set 95114_PM_s_at - -
          Murine Genome U74v2 Set 95114_s_at - -
          Murine Genome U74v2 Set 95113_PM_at - -
          Mouse Genome 430A 2.0 Array 1451387_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451387_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451387_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451387_s_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3520133 - -
          Illumina mouse-6 v1.0 expression beadchip 3520133 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003520133 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2669540 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2669540 - -
          mouse-6 v1.1 (Illumina) scl50143.3.1_18-S - -
          Illumina Sentrix mouse-6.v1 bead array 3520133 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl50143.3.1_18-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3520133 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3041584 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2669540 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3116330 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2669540 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3041584 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3116330 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl50143.3.1_18-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0