Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503255_s1
          See other FAM122A GE Assays ›
          Gene Symbol:
          Fam122a
          Gene Name:
          family with sequence similarity 122, member A
          Gene Aliases:
          2410124L17Rik, 2900009I07Rik, ENSMUSG00000051082, Gm9849
          Chromosome Location:
          Chr.19: 24475779 - 24477474 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503255_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam122a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026520.3 NP_080796.1 1 366 56
          GenBank mRNA AK005219.1 - 1 356 56
          AK010777.1 - 1 378 56
          AK011741.1 - 1 350 56
          AK033646.1 - 1 366 56
          AK077654.1 - 1 348 56
          AK172089.1 - 1 351 56
          BC028637.1 - 1 334 1248203 56
          BC083062.1 - 1 356 5361132 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1900001282.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam122a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023816 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023818 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023819 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023820 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK077654.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK172089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 BC028637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 BC083062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK011741.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK077654.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK172089.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 BC028637.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 BC083062.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P396811 AK011741.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141099 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK077654.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK172089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 BC028637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 BC083062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK011741.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK077654.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK172089.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 BC028637.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 BC083062.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P396811 AK011741.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023819 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023841 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023825 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023815 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023832 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023816 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023842 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023818 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023806 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023839 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023857 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023809 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023833 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023855 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023823 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023801 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023800 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023802 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023803 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32023824 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141099 - -
          Whole Mouse Genome G4122A A_51_P396811 AK077654.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 AK172089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 BC028637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 BC083062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 AK011741.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 AK077654.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 AK172089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 BC028637.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 BC083062.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P396811 AK011741.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132223 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106007 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141099 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14486756 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14299020 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14299016 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14299019 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023821 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023822 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023816 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023819 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023817 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023820 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P32023818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023867 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023850 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023881 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32023853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023811 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023834 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14225671 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023822 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023859 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14429800 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023850 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14272789 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023849 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023843 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023867 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023806 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023842 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14141080 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023855 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14276015 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023845 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023860 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14225667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023847 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023853 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023836 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023809 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023831 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023844 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023827 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023878 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023825 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14152760 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023839 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14326688 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023877 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32023875 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10466797 - -
          Affymetrix Mouse Gene 1.0 ST Array 10462131 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466796 - -
          Affymetrix Mouse Gene 1.0 ST Array 10466795 - -
          Affymetrix HT MG-430 PM Array Plate 1453981_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1450411_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1430769_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10462130 - -
          Affymetrix Mouse Gene 1.1 ST Array 10466794 - -
          Mouse Genome 430 2.0 Array 1430769_PM_s_at - -
          Mouse Genome 430 2.0 Array 1453981_PM_at - -
          Mouse Genome 430 2.0 Array 1453981_at - -
          Mouse Genome 430 2.0 Array 1450411_PM_at - -
          Mouse Genome 430 2.0 Array 1430769_s_at - -
          Mouse Genome 430 2.0 Array 1450411_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68034_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68034_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1900011575.mm.1 TC1900001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011575.mm.1 NM_026520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011575.mm.1 NM_026520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011576.mm.1 TC1900001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011576.mm.1 NM_026520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011576.mm.1 NM_026520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011577.mm.1 TC1900001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011577.mm.1 NM_026520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011577.mm.1 NM_026520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011579.mm.1 TC1900001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011579.mm.1 NM_026520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011579.mm.1 NM_026520.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011578.mm.1 TC1900001282.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1900011578.mm.1 NM_026520.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1900011578.mm.1 NM_026520.3 Refseq
          Mouse Expression Set 430 1453981_at - -
          Mouse Expression Set 430 1430769_s_at - -
          Mouse Expression Set 430 1453981_PM_at - -
          Mouse Expression Set 430 1430769_PM_s_at - -
          Mouse Expression Set 430 1450411_at - -
          Mouse Expression Set 430 1450411_PM_at - -
          Mouse Genome 430A 2.0 Array 1450411_at - -
          Mouse Genome 430A 2.0 Array 1453981_at - -
          Mouse Genome 430A 2.0 Array 1430769_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1430769_s_at - -
          Mouse Genome 430A 2.0 Array 1450411_PM_at - -
          Mouse Genome 430A 2.0 Array 1453981_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453981_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450411_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1450411_at - -
          Affymetrix HT Mouse Genome 430A Array 1453981_at - -
          Affymetrix HT Mouse Genome 430A Array 1430769_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1430769_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 752144 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6040242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2668661 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2668661 - -
          mouse-6 v1.1 (Illumina) scl52635.1.33_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl52635.1.33_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2668661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2668661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6c59565f7c-q2rrq:80/100.66.130.62:80.
          git-commit: 653a2953ec6bb2e3dd1e37289b36fc4e0ab15bfe
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-Offline