Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00503776_g1
          See other IVNS1ABP GE Assays ›
          Gene Symbol:
          Ivns1abp
          Gene Name:
          influenza virus NS1A binding protein
          Gene Aliases:
          1190004M08Rik, 1700126I16Rik, AA960440, HSPC068, ND1, NS-1, NS1-BP, Nd1-L, Nd1-S, mKIAA0850
          Chromosome Location:
          Chr.1: 151344308 - 151364445 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00503776_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ivns1abp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001039511.1 NP_001034600.1 4 - 5 788 108
          NM_001039512.1 NP_001034601.1 4 - 5 788 108
          NM_054102.2 NP_473443.2 4 - 5 788 108
          GenBank mRNA AB055737.1 - 4 - 5 784 108
          AB055738.1 - 4 - 5 784 108
          AK007291.1 - 4 - 5 738 108
          AK129229.1 - 4 - 5 726 108
          AK144619.1 - 4 - 5 767 108
          AK144835.1 - 4 - 5 769 108
          AK145071.1 - 4 - 5 779 108
          AK145845.1 - 4 - 5 738 108
          AK146001.1 - 4 - 5 769 108
          AK146329.1 - 1 - 2 112 108
          AK146494.1 - 4 - 5 741 108
          AK146645.1 - 4 - 5 739 108
          AK159254.1 - 4 - 5 770 108
          AK179875.1 - 3 - 4 340 108
          AK181493.1 - 3 - 4 365 108
          BC004092.1 - 3 - 4 488 3593827 108
          BC040250.1 - 4 - 5 780 108
          DQ914522.1 - 4 - 5 740 108
          DQ914523.1 - 4 - 5 710 108

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100001351.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the DQ914523.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ivns1abp

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of intrinsic apoptotic signaling pathway
          protein binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 TC0100001351.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_001039511.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_001039511.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_001039512.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_001039512.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_054102.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100011184.mm.1 NM_054102.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0