Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504027_s1
          See other KTI12 GE Assays ›
          Gene Symbol:
          Kti12 A730015C16Rik
          Gene Name:
          KTI12 homolog, chromatin associated (S. cerevisiae)
          RIKEN cDNA A730015C16 gene
          Gene Aliases:
          1110001A12Rik, AI326810, AU014758
          ENSMUSG00000053880
          Chromosome Location:
          Chr.4: 108847857 - 108849412 on Build GRCm38
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          Assay ID Mm00504027_s1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kti12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029571.2 NP_083847.1 1 149 70
          GenBank mRNA AK003197.1 - 1 150 70
          AK148185.1 - 1 139 70
          AK150178.1 - 1 137 70
          AK156755.1 - 1 117 70
          BC021493.1 - 1 232 5345710 70
          Gene Symbol: A730015C16Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK042682.1 - 1 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400001158.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kti12
          Click here to find siRNAs for Gene A730015C16Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          nucleotide binding
          molecular_function
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197992 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197990 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197993 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK148185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK150178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK156755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 BC021493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 NM_029571.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK148185.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK150178.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 AK156755.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 BC021493.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P394728 NM_029571.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155529 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK148185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK150178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK156755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 BC021493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 NM_029571.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK148185.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK150178.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 AK156755.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 BC021493.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P394728 NM_029571.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197982 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197996 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197993 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23198000 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197970 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197963 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197987 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197938 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197975 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197976 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197990 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197983 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197994 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197978 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197988 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197974 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197961 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197998 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197997 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197941 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197986 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197985 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197999 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23197992 - -
          Whole Mouse Genome G4122A A_51_P394728 AK148185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 AK150178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 AK156755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 BC021493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 NM_029571.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 AK148185.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 AK150178.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 AK156755.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 BC021493.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P394728 NM_029571.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P133127 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647842 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647812 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647837 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03647826 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197989 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197992 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23197990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197992 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197990 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23197997 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03527743 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197983 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04056166 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04060960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03610499 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03647843 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03217273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197941 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197985 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23198000 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197947 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197950 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197974 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197982 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197996 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197976 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197949 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197939 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03217272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197987 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197999 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03840859 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03647819 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197960 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23197968 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04060964 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10506882 - -
          Affymetrix Mouse Gene 1.0 ST Array 10506881 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515050 - -
          Affymetrix HT MG-430 PM Array Plate 1416106_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10506880 - -
          Affymetrix Mouse Gene 1.1 ST Array 10515049 - -
          Mouse Genome 430 2.0 Array 1416106_at - -
          Mouse Genome 430 2.0 Array 1416106_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100087_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100087_at - -
          Affymetrix Mouse Clariom S Array PSR0400008282.mm.1 TC0400001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008283.mm.1 TC0400001158.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400008283.mm.1 NM_029571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400008283.mm.1 NM_029571.2 Refseq
          Mouse Expression Set 430 1416106_at - -
          Mouse Expression Set 430 1416106_PM_at - -
          Mouse Genome 430A 2.0 Array 1416106_at - -
          Mouse Genome 430A 2.0 Array 1416106_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1416106_at - -
          Affymetrix HT Mouse Genome 430A Array 1416106_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 569769 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599412 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599412 - -
          mouse-6 v1.1 (Illumina) scl25138.1_25-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl25138.1_25-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599412 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599412 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl25138.1_25-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0