Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00504795_g1
          See other PRSS8 GE Assays ›
          Gene Symbol:
          Prss8
          Gene Name:
          protease, serine 8 (prostasin)
          Gene Aliases:
          2410039E18Rik, AI313909, C79772, CAP1, fr, mCAP1
          Chromosome Location:
          Chr.7: 127925717 - 127930142 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00504795_g1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prss8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133351.3 NP_579929.1 5 - 6 935 69
          GenBank mRNA AB038244.1 - 5 - 6 916 69
          AF188613.1 - 5 - 6 849 69
          AF378085.2 - 5 - 6 926 69
          AK078696.1 - 5 - 6 890 69
          AY335911.1 - 6 - 7 783 69
          BC003851.1 - 5 - 6 909 3600399 69

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700004346.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prss8

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          proteolysis
          positive regulation of sodium ion transport
          transepithelial transport
          serine-type endopeptidase activity
          peptidase activity
          serine-type peptidase activity
          hydrolase activity
          sodium channel regulator activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508221 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508223 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071191 AK010640.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071191 AK078696.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071191 AY335911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071191 BC003851.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2071191 AF188613.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013586 AY335911.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013586 AF378085.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013586 AB038244.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013586 NM_133351.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2013586 AK018002.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 455345 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 126643 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P154663 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071191 AK010640.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071191 AK078696.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071191 AY335911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071191 BC003851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2071191 AF188613.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013586 AY335911.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013586 AF378085.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013586 AB038244.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013586 NM_133351.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2013586 AK018002.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P410703 AB038244.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508177 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508186 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508213 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508205 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508189 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508168 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508219 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508198 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508174 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508173 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25508197 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P154663 - -
          Whole Mouse Genome G4122A A_51_P410703 AB038244.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137720 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P154663 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632934 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632928 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632938 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632926 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632933 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06632925 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508219 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508220 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508218 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508223 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508222 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25508221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508098 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508168 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508115 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25508090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06440925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508168 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508181 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06632935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508152 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06632936 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06632925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05978610 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06313703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06413702 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25508163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568338 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568336 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568337 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568333 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568335 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568334 - -
          Affymetrix HT MG-430 PM Array Plate 1418320_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568332 - -
          Affymetrix Mouse Gene 1.1 ST Array 10557831 - -
          Mouse Genome 430 2.0 Array 1418320_PM_at - -
          Mouse Genome 430 2.0 Array 1418320_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76560_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 76560_at - -
          Affymetrix Mouse Clariom S Array PSR0700036515.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036515.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036515.mm.1 NM_133351.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036516.mm.1 TC0700004346.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700036516.mm.1 NM_133351.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700036516.mm.1 NM_133351.3 Refseq
          Mouse Expression Set 430 1418320_at - -
          Mouse Expression Set 430 1418320_PM_at - -
          Murine Genome U74v2 Set 100909_at - -
          Murine Genome U74v2 Set 160566_at - -
          Murine Genome U74v2 Set 160566_PM_at - -
          Murine Genome U74v2 Set 100909_PM_at - -
          Mouse Genome 430A 2.0 Array 1418320_at - -
          Mouse Genome 430A 2.0 Array 1418320_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418320_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1418320_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170577 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170577 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2740966 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2740966 - -
          mouse-6 v1.1 (Illumina) scl30628.6_19-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170577 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30628.6_19-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170577 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2846148 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2740965 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2740965 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846148 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0