Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00505427_g1
          See other PLA2G15 GE Assays ›
          Gene Symbol:
          Pla2g15
          Gene Name:
          phospholipase A2, group XV
          Gene Aliases:
          ACS, C87498, LLPL, Lpla2, Lypla3
          Chromosome Location:
          Chr.8: 106150351 - 106164715 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00505427_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pla2g15
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133792.2 NP_598553.1 3 - 4 493 64
          GenBank mRNA AF468958.1 - 3 - 4 405 64
          AK028614.1 - 3 - 4 493 64
          AK085194.1 - 3 - 4 482 64
          AK163111.1 - 3 - 4 482 64
          AK170814.1 - 3 - 4 481 64
          BC019373.1 - 3 - 4 493 3964729 64

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001244.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pla2g15

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid metabolic process
          ceramide metabolic process
          phosphatidylcholine metabolic process
          dihydrolipoamide branched chain acyltransferase activity
          lysophospholipase activity
          sterol O-acyltransferase activity
          N-acetyltransferase activity
          O-acyltransferase activity
          palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
          carnitine O-acyltransferase activity
          acetyltransferase activity
          C-acyltransferase activity
          palmitoyltransferase activity
          N-acyltransferase activity
          acylglycerol O-acyltransferase activity
          serine O-acyltransferase activity
          O-acetyltransferase activity
          O-octanoyltransferase activity
          octanoyltransferase activity
          O-palmitoyltransferase activity
          S-acyltransferase activity
          S-acetyltransferase activity
          S-malonyltransferase activity
          malonyltransferase activity
          C-palmitoyltransferase activity
          transferase activity
          acyltransferase activity
          succinyltransferase activity
          N-succinyltransferase activity
          O-succinyltransferase activity
          S-succinyltransferase activity
          sinapoyltransferase activity
          O-sinapoyltransferase activity
          hydrolase activity
          peptidyl-lysine N6-myristoyltransferase activity
          peptidyl-lysine N6-palmitoyltransferase activity
          benzoyl acetate-CoA thiolase activity
          3-hydroxybutyryl-CoA thiolase activity
          3-ketopimelyl-CoA thiolase activity
          N-palmitoyltransferase activity
          myristoyltransferase activity
          acyl-CoA N-acyltransferase activity
          protein-cysteine S-myristoyltransferase activity
          protein-cysteine S-acyltransferase activity
          dihydrolipoamide S-acyltransferase activity
          glucosaminyl-phosphatidylinositol O-acyltransferase activity
          ergosterol O-acyltransferase activity
          lanosterol O-acyltransferase activity
          naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
          2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
          2-methylhexanoyl-CoA C-acetyltransferase activity
          butyryl-CoA 2-C-propionyltransferase activity
          2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
          alpha-aminoadipate acetyltransferase activity
          keto acid formate lyase activity
          Ras palmitoyltransferase activity
          azetidine-2-carboxylic acid acetyltransferase activity
          calcium-independent phospholipase A2 activity
          peptidyl-lysine acetyltransferase activity
          acetyl-CoA:L-lysine N6-acetyltransferase
          UDP-3-O-(R-3-hydroxymyristoyl)-glucosamine N-acyltransferase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800009972.mm.1 TC0800001244.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009972.mm.1 NM_133792.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009972.mm.1 NM_133792.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0