Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508298_m1
          See other MTERF4 GE Assays ›
          Gene Symbol:
          Mterf4
          Gene Name:
          mitochondrial transcription termination factor 4
          Gene Aliases:
          1810059A23Rik, 4933412H03Rik, Mterfd2
          Chromosome Location:
          Chr.1: 93299211 - 93305965 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508298_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mterf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178051.3 NP_835152.1 2 - 3 541 87
          GenBank mRNA AK077142.1 - 2 - 3 541 87
          AK079098.1 - 2 - 3 533 87
          BC002280.1 - 1 - 2 154 3497421 87
          BC055877.1 - 1 - 2 485 4236729 87
          BC064709.1 - 2 - 3 534 30314353 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0100002831.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mterf4

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          rRNA processing
          mitochondrial transcription
          protein targeting to mitochondrion
          heart development
          ribosome assembly
          camera-type eye development
          double-stranded DNA binding
          RNA binding
          rRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454695 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454693 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454692 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC055877.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC064709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC002280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 AK077142.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 AK079098.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC055877.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC064709.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 BC002280.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 AK077142.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P69756 AK079098.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 60908 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 867926 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 482413 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141197 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC055877.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC064709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC002280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 AK077142.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 AK079098.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC055877.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC064709.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 BC002280.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 AK077142.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P69756 AK079098.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454715 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454675 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454685 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454682 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454673 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454672 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454687 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454711 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454680 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454708 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20454703 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141197 - -
          Whole Mouse Genome G4122A A_51_P208701 AK079098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P208701 NM_178051.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC055877.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC064709.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC002280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 AK077142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 AK079098.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC055877.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC064709.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 BC002280.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 AK077142.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P69756 AK079098.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126865 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623564 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623555 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623568 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623561 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623571 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623558 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00623547 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00345841 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454693 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454695 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454694 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P20454692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454675 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20454625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623546 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00239841 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00565894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623543 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00345845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00239840 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00565902 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00565905 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00565889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00569313 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623558 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00565896 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00783889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623555 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623557 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623554 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454658 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00601945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454709 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00601948 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00623552 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20454652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00699235 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10356773 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356772 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356777 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356776 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356775 - -
          Affymetrix Mouse Gene 1.0 ST Array 10356774 - -
          Affymetrix HT MG-430 PM Array Plate 1424430_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10356771 - -
          Mouse Genome 430 2.0 Array 1458808_PM_at - -
          Mouse Genome 430 2.0 Array 1458808_at - -
          Mouse Genome 430 2.0 Array 1424430_at - -
          Mouse Genome 430 2.0 Array 1424430_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69821_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69821_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100022924.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022924.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022919.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022919.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022922.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022922.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022911.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022911.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022913.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022913.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022914.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022914.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022915.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022915.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022921.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022921.mm.1 NM_178051.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0100022916.mm.1 TC0100002831.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100022916.mm.1 NM_178051.4 Refseq
          Mouse Expression Set 430 1424430_at - -
          Mouse Expression Set 430 1424430_PM_at - -
          Mouse Genome 430A 2.0 Array 1424430_PM_at - -
          Mouse Genome 430A 2.0 Array 1424430_at - -
          Affymetrix HT Mouse Genome 430A Array 1424430_at - -
          Affymetrix HT Mouse Genome 430A Array 1424430_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 673058 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1570333 - -
          Illumina mouse-6 v1.0 expression beadchip 1570333 - -
          Illumina mouse-6 v1.0 expression beadchip 4540139 - -
          Illumina mouse-6 v1.0 expression beadchip 4230176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004540139 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230176 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570333 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218949 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1236049 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2679695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2679695 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218949 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236049 - -
          mouse-6 v1.1 (Illumina) scl0069821.2_297-S - -
          mouse-6 v1.1 (Illumina) scl16462.4_619-S - -
          mouse-6 v1.1 (Illumina) scl000856.1_265-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570333 - -
          Illumina Sentrix mouse-6.v1 bead array 4230176 - -
          Illumina Sentrix mouse-6.v1 bead array 4540139 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000856.1_265-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0069821.2_297-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl16462.4_619-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4540139 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230176 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2679695 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1218949 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1236049 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218949 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2679695 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236049 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000856.1_265-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl16462.4_619-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0069821.2_297-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6467d75d44-xp7n2:80/100.66.130.62:80.
          git-commit: 8ea1bebb48d33b8b7c0e2103582aa482e6db8664
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0