Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00508455_m1
          See other SPC25 GE Assays ›
          Gene Symbol:
          Spc25
          Gene Name:
          SPC25, NDC80 kinetochore complex component, homolog (S. cerevisiae)
          Gene Aliases:
          2600017H08Rik, 2610205L13Rik, Spbc25
          Chromosome Location:
          Chr.2: 69193895 - 69206215 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00508455_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Spc25
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK003408.1 - 5 - 6 513 96
          AK011241.1 - 6 - 7 96
          AK146727.1 - 6 - 7 1141 96
          AK151554.1 - 6 - 7 672 96
          BC027121.1 - 6 - 7 639 5359960 96
          BC033605.1 - 6 - 7 888 5369405 96
          CX222514.1 - 6 - 7 698 96

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003697.mm.1

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307667 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307668 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965154 BC027121.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965154 AK146727.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965154 AK151554.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965154 BC033605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1965154 AK011241.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626054 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 463257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 113537 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 909329 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511146 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862314 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AF421882.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AJ310548.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AK011502.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AK044787.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AK165972.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 AK170732.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 BC044926.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 BC058278.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 465788 NM_028133.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 567733 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715952 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 155889 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102013 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P115347 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965154 BC027121.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965154 AK146727.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965154 AK151554.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965154 BC033605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1965154 AK011241.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307714 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307718 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307677 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307661 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307674 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307676 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307679 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307681 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307671 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307670 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307697 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21307663 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00396219 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102013 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P115347 - -
          Whole Mouse Genome G4122A A_51_P514700 AK011241.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132148 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P115347 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102013 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01308922 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01308919 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02077219 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01308915 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01156449 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01308926 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307670 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307668 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307672 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21307667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307588 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307647 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21307694 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02192158 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307659 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02066109 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307664 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01749592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307697 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01308916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307677 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307604 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01338393 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307641 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307701 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307611 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307686 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307608 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01156449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307647 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307633 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01308924 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307644 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307606 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02033084 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02275297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307639 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307704 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01170685 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307635 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01308915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01494553 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01484612 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307605 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21307631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01296700 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10483402 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483404 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483405 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483407 - -
          Affymetrix Mouse Gene 1.0 ST Array 10483403 - -
          Affymetrix HT MG-430 PM Array Plate 1453815_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1424118_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10483401 - -
          Mouse Genome 430 2.0 Array 1453815_at - -
          Mouse Genome 430 2.0 Array 1424118_PM_a_at - -
          Mouse Genome 430 2.0 Array 1424118_a_at - -
          Mouse Genome 430 2.0 Array 1453815_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66442_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 66442_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200031533.mm.1 TC0200003697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031533.mm.1 NM_001199123.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031533.mm.1 NM_001199124.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031533.mm.1 NM_025565.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031530.mm.1 TC0200003697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031530.mm.1 NM_001199123.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031530.mm.1 NM_001199124.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031530.mm.1 NM_025565.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031532.mm.1 TC0200003697.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200031532.mm.1 NM_001199123.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031532.mm.1 NM_001199124.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200031532.mm.1 NM_025565.4 Refseq
          Mouse Expression Set 430 1424118_PM_a_at - -
          Mouse Expression Set 430 1424118_a_at - -
          Mouse Genome 430A 2.0 Array 1424118_PM_a_at - -
          Mouse Genome 430A 2.0 Array 1424118_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453815_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424118_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1453815_at - -
          Affymetrix HT Mouse Genome 430A Array 1424118_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 742968 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6860242 - -
          Illumina mouse-6 v1.0 expression beadchip 6860242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006860242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1255960 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1255960 - -
          mouse-6 v1.1 (Illumina) scl19174.6.1_6-S - -
          Illumina Sentrix mouse-6.v1 bead array 6860242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19174.6.1_6-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2901180 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1255960 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1255960 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2901180 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0