Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam92a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026558.4 | NP_080834.3 | 1 - 2 | 318 | 61 | |
GenBank mRNA | AK013129.1 | - | 1 - 2 | 149 | 61 | |
AK019574.1 | - | 1 - 2 | 146 | 61 | ||
AK032690.1 | - | 1 - 2 | 298 | 61 | ||
AK078438.1 | - | 1 - 2 | 299 | 61 | ||
AK088705.1 | - | 1 - 2 | 299 | 61 | ||
BC020162.1 | - | 112 | 3991990 | 61 | ||
BC055848.1 | - | 1 - 2 | 128 | 61 | ||
CJ064072.1 | - | 1 - 2 | 319 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |