Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509660_m1
          See other LHFPL5 GE Assays ›
          Gene Symbol:
          Lhfpl5
          Gene Name:
          lipoma HMGIC fusion partner-like 5
          Gene Aliases:
          9330179O15Rik, Tmhs, hscy
          Chromosome Location:
          Chr.17: 28575371 - 28583593 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509660_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Lhfpl5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026571.2 NP_080847.2 1 - 2 698 86
          GenBank mRNA AK020670.1 - 1 - 2 686 86
          BC099483.1 - 1 - 2 596 30848018 86
          BC125264.1 - 1 - 2 535 86
          BC147842.1 - 1 - 2 496 9087559 86
          BC147847.1 - 1 - 2 496 9087564 86

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1700000483.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Lhfpl5

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          ion transport
          sensory perception of sound
          detection of mechanical stimulus involved in sensory perception of sound
          auditory receptor cell stereocilium organization
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123145 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123146 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127433 AK020389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127433 BC099483.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2127433 NM_026571.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 610333 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 740408 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 711302 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481583 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 783508 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117222 BC147842.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2117222 BC147847.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127433 AK020389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127433 BC099483.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2127433 NM_026571.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P406193 AK020389.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123132 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123144 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123155 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123140 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123161 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123149 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31123152 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08751130 - -
          Whole Mouse Genome G4122A A_51_P406193 AK020389.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08751130 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376325 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376321 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376311 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376320 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13376319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123144 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123146 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31123145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123144 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123195 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123170 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123148 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31123173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123154 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13489398 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13569032 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13376313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13376323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123129 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123125 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123177 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13111667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123141 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123130 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123147 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123126 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123162 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13256259 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123189 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123187 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437724 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123140 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123142 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123173 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123169 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13437722 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123170 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123164 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123184 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123150 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13569019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123146 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123152 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123132 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123159 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31123165 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10443389 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443385 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443387 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443384 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443386 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443390 - -
          Affymetrix Mouse Gene 1.0 ST Array 10443388 - -
          Affymetrix HT MG-430 PM Array Plate 1429266_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10443383 - -
          Mouse Genome 430 2.0 Array 1429266_PM_at - -
          Mouse Genome 430 2.0 Array 1429266_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328789_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 328789_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700004187.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004187.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004187.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004186.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004186.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004186.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004189.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004189.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004189.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004192.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004192.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004192.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004188.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004188.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004188.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004193.mm.1 TC1700000483.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700004193.mm.1 NM_026571.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004193.mm.1 NM_026571.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1700004190.mm.1 TC1700000483.mm.1 MTA TC
          Mouse Expression Set 430 1429266_PM_at - -
          Mouse Expression Set 430 1429266_at - -
          Affymetrix HT Mouse Genome 430B Array 1429266_at - -
          Affymetrix HT Mouse Genome 430B Array 1429266_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 691260 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2518178 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2518178 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6b89786976-jw7rs:80/100.66.135.20:80.
          git-commit: 3f48773a9a35f89d7dca75e8633f1a0b0cdc18d9
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0