Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00509772_g1
          See other FAM166A GE Assays ›
          Gene Symbol:
          Fam166a
          Gene Name:
          family with sequence similarity 166, member A
          Gene Aliases:
          4931415M17
          Chromosome Location:
          Chr.2: 25214724 - 25222281 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00509772_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam166a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026624.3 NP_080900.1 6 - 7 929 70
          GenBank mRNA AK016459.1 - 6 - 7 931 70
          BC061039.1 - 6 - 7 932 6704633 70
          BX631039.1 - 1 - 2 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200000401.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fam166a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 BX631039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 NM_026624.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 AK016459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 BC061039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 BX631039.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 NM_026624.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 AK016459.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P541258 BC061039.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 580059 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 123896 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 5497 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 BX631039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 NM_026624.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 AK016459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 BC061039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 BX631039.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 NM_026624.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 AK016459.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P541258 BC061039.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070609 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070607 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070606 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070616 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070610 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070621 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04456865 - -
          Whole Mouse Genome G4122A A_52_P541258 BX631039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 NM_026624.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 AK016459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 BC061039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 BX631039.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 NM_026624.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 AK016459.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P541258 BC061039.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P107912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01885004 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01885006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070577 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070556 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01293595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01604643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070603 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070616 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070595 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070614 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070599 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01238286 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070601 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070630 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070609 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070619 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01885004 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070594 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070625 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070596 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01653150 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01653140 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070606 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469948 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469950 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469946 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469947 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469945 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469944 - -
          Affymetrix HT MG-430 PM Array Plate 1453026_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469943 - -
          Mouse Genome 430 2.0 Array 1453026_at - -
          Mouse Genome 430 2.0 Array 1453026_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68222_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68222_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200002213.mm.1 TC0200000401.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200002213.mm.1 NM_026624.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002213.mm.1 NM_026624.3 Refseq
          Mouse Expression Set 430 1453026_at - -
          Mouse Expression Set 430 1453026_PM_at - -
          Mouse Genome 430A 2.0 Array 1453026_at - -
          Mouse Genome 430A 2.0 Array 1453026_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1453026_at - -
          Affymetrix HT Mouse Genome 430A Array 1453026_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 799781 - -
          ABI Mouse Genome Survey Microarray 855301 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 5420056 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005420056 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2622410 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2622410 - -
          mouse-6 v1.1 (Illumina) scl21189.7.1_27-S - -
          Illumina Sentrix mouse-6.v1 bead array 5420056 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21189.7.1_27-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5420056 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1234184 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234184 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0