Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00510091_m1
          See other RWDD2A GE Assays ›
          Gene Symbol:
          Rwdd2a
          Gene Name:
          RWD domain containing 2A
          Gene Aliases:
          1700030C20Rik, AI848608, Rwdd2
          Chromosome Location:
          Chr.9: 86571988 - 86574899 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00510091_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rwdd2a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027100.2 NP_081376.2 1 - 2 164 83
          GenBank mRNA AK006533.1 - 1 - 2 163 83
          BC115691.1 - 1 - 2 164 40104313 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0900001115.mm.1

          Additional Information:

          This assay is in the 5'UTR region of the AK006533.1, BC115691.1, NM_027100.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rwdd2a

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700015 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 AK006533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_001145969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 AK006533.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115691.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 BC115692.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_001145969.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563284 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P165096 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 AK006533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_001145969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_027100.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 AK006533.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115691.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 BC115692.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_001145969.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700018 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700015 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700021 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700014 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26700017 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P165096 - -
          Whole Mouse Genome G4122A A_52_P554436 AK006533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_001145969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_027100.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 AK006533.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115691.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 BC115692.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_001145969.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P554436 NM_027100.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137626 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P165096 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779072 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779077 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07779078 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700014 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26700015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26700021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07843403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07965323 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779077 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700021 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700051 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07965310 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700023 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700024 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700046 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07991571 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07779078 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26700053 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10587608 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587606 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587605 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587607 - -
          Affymetrix Mouse Gene 1.0 ST Array 10587609 - -
          Affymetrix HT MG-430 PM Array Plate 1428963_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10587604 - -
          Mouse Genome 430 2.0 Array 1428963_PM_at - -
          Mouse Genome 430 2.0 Array 1428963_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69519_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69519_at - -
          Affymetrix Mouse Clariom S Array PSR0900008776.mm.1 TC0900001115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008776.mm.1 NM_001145968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008776.mm.1 NM_001145968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008776.mm.1 NM_027100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008776.mm.1 NM_027100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 TC0900001115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_001145968.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_001145968.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_001145969.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_001145969.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_027100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008778.mm.1 NM_027100.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008777.mm.1 TC0900001115.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900008777.mm.1 NM_027100.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900008777.mm.1 NM_027100.2 Refseq
          Mouse Expression Set 430 1428963_PM_at - -
          Mouse Expression Set 430 1428963_at - -
          Affymetrix HT Mouse Genome 430B Array 1428963_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428963_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 310349 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 1660079 - -
          Illumina mouse-6 v1.0 expression beadchip 1660079 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001660079 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2774528 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2774528 - -
          mouse-6 v1.1 (Illumina) scl36646.3.1_24-S - -
          Illumina Sentrix mouse-6.v1 bead array 1660079 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl36646.3.1_24-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1660079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2774528 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl36646.3.1_24-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-65cb46898d-nlgdw:80/100.66.128.131:80.
          git-commit: e5493f66f4597fc66e516c8a379ca3b6bed48f5b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0