Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511030_m1
          See other ZSWIM1 GE Assays ›
          Gene Symbol:
          Zswim1
          Gene Name:
          zinc finger SWIM-type containing 1
          Gene Aliases:
          2410003H12Rik, AI850991, BB046916
          Chromosome Location:
          Chr.2: 164822686 - 164826867 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511030_m1
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zswim1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028028.2 NP_082304.1 1 - 2 516 106
          GenBank mRNA AK010358.1 - 1 - 2 190 106
          AK149816.1 - 1 - 2 190 106
          AK150174.1 - 1 - 2 190 106
          BC046831.1 - 1 - 2 516 6812759 106

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK010358.1, BC046831.1, NM_028028.2 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zswim1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819954 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 BC046831.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK150174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK197876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 BC025184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 BC046831.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK150174.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK197876.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 BC025184.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 379668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 197011 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348331 AK082020.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348331 AK170159.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348331 AK171246.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348331 BC010462.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 348331 NM_145373.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111233 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 144593 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 BC046831.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK150174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK197876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 BC025184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK010358.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 BC046831.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK150174.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK197876.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 BC025184.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 BC046831.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK150174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK197876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 BC025184.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 BC046831.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK150174.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK197876.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 BC025184.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819969 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21819956 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04773095 - -
          Whole Mouse Genome G4122A A_51_P232474 BC046831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK150174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK197876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 BC025184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK010358.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 BC046831.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK150174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK197876.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 BC025184.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P232474 AK010358.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126734 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01643178 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02159823 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01643179 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21819954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819976 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819967 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819969 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21819972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21820028 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01608095 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819977 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819973 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01622850 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01622855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819965 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212408 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02159822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819955 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01622838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01622848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21819984 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01643185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02159823 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02227382 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01643179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01212417 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10478593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10478592 - -
          Affymetrix HT MG-430 PM Array Plate 1451189_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10478590 - -
          Mouse Genome 430 2.0 Array 1451189_PM_at - -
          Mouse Genome 430 2.0 Array 1451189_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71971_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71971_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200020727.mm.1 TC0200002536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020727.mm.1 NM_028028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020727.mm.1 NM_028028.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020726.mm.1 TC0200002536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020726.mm.1 NM_028028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020726.mm.1 NM_028028.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020730.mm.1 TC0200002536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020730.mm.1 NM_028028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020730.mm.1 NM_028028.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020731.mm.1 TC0200002536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020731.mm.1 NM_028028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020731.mm.1 NM_028028.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020729.mm.1 TC0200002536.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200020729.mm.1 NM_028028.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200020729.mm.1 NM_028028.2 Refseq
          Mouse Expression Set 430 1451189_at - -
          Mouse Expression Set 430 1451189_PM_at - -
          Mouse Genome 430A 2.0 Array 1451189_at - -
          Mouse Genome 430A 2.0 Array 1451189_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451189_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451189_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 593436 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 7050239 - -
          Illumina mouse-6 v1.0 expression beadchip 7050239 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0007050239 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1245683 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1245683 - -
          mouse-6 v1.1 (Illumina) scl19923.3_605-S - -
          Illumina Sentrix mouse-6.v1 bead array 7050239 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl19923.3_605-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 7050239 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2846047 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2846047 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl19923.3_605-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0