Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm00511334_g1
          See other STRADA GE Assays ›
          Gene Symbol:
          Strada
          Gene Name:
          STE20-related kinase adaptor alpha
          Gene Aliases:
          2610019A05Rik, 6030402H20Rik, AI480680, E130112C09Rik
          Chromosome Location:
          Chr.11: 106162974 - 106193604 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm00511334_g1
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Strada
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001252448.1 NP_001239377.1 11 - 12 1282 85
          NM_028126.3 NP_082402.1 9 - 10 1171 85
          GenBank mRNA AK011451.1 - 9 - 10 1117 85
          AK138358.1 - 11 - 12 1283 85
          AY341872.1 - 9 - 10 1130 85
          BC052063.1 - 1178 85
          BC058517.1 - 9 - 10 1072 5717255 85

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004297.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Strada

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          protein export from nucleus
          cell cycle
          Rho protein signal transduction
          regulation of mitotic cell cycle
          cell migration
          actin cytoskeleton organization
          stress-activated protein kinase signaling cascade
          activation of protein kinase activity
          regulation of apoptotic process
          regulation of MAPK cascade
          protein heterooligomerization
          positive regulation of protein serine/threonine kinase activity
          protein kinase activity
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          transferase activity, transferring phosphorus-containing groups
          kinase binding
          protein kinase activator activity
          protein serine/threonine kinase activator activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100036570.mm.1 TC1100004297.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100036570.mm.1 NM_001252448.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036570.mm.1 NM_001252448.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036570.mm.1 NM_028126.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100036570.mm.1 NM_028126.3 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d58dc5f8-fsfvr:80/100.66.134.75:80.
          git-commit: 38b8a393e477ae4d9d89cc5f7a29948d21ae29e2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc